Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89856168-89856393 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPIRE2
Entrez Gene ID
84501
Gene Name
spire type actin nucleation factor 2
Gene Aliases
Spir-2
UniGene
Hs.461786
Gene Chromosome Location
Chr.16: 89817477-89871319 on Build GRCh38
Transcripts
NM_032451.1 XM_011523379.1 XM_011523380.2 XM_011523381.1 XM_011523384.1 XM_011523385.1 XM_017023779.1 XM_017023780.1 XM_017023781.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5084786 c.1100G>A p.R367H Chr.16: 89856234-89856234 on Build GRCh38
6380655 c.1076T>C p.V359A Chr.16: 89856210-89856210 on Build GRCh38
974979 c.1083C>T p.G361G Chr.16: 89856217-89856217 on Build GRCh38
4063668 c.1090T>G p.W364G Chr.16: 89856224-89856224 on Build GRCh38
1227401 c.1070G>A p.R357H Chr.16: 89856204-89856204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753893073 Chr.16: 89856193-89856193 on Build GRCh38
rs552615145 Chr.16: 89856237-89856237 on Build GRCh38
rs573573061 Chr.16: 89856195-89856195 on Build GRCh38
rs909201402 Chr.16: 89856263-89856263 on Build GRCh38
rs767480736 Chr.16: 89856218-89856218 on Build GRCh38
rs770539114 Chr.16: 89856213-89856213 on Build GRCh38
rs772218352 Chr.16: 89856341-89856341 on Build GRCh38
rs762472678 Chr.16: 89856212-89856212 on Build GRCh38
rs753887387 Chr.16: 89856235-89856235 on Build GRCh38
rs550480688 Chr.16: 89856282-89856282 on Build GRCh38
rs778934706 Chr.16: 89856261-89856261 on Build GRCh38
rs113325862 Chr.16: 89856319-89856319 on Build GRCh38
rs752486759 Chr.16: 89856219-89856219 on Build GRCh38
rs373707880 Chr.16: 89856234-89856234 on Build GRCh38
rs573677603 Chr.16: 89856300-89856300 on Build GRCh38
rs1048653383 Chr.16: 89856288-89856288 on Build GRCh38
rs772628067 Chr.16: 89856209-89856209 on Build GRCh38
rs780381068 Chr.16: 89856207-89856207 on Build GRCh38
rs116983808 Chr.16: 89856327-89856327 on Build GRCh38
rs759260212 Chr.16: 89856217-89856217 on Build GRCh38
rs757409105 Chr.16: 89856236-89856236 on Build GRCh38
rs772468359 Chr.16: 89856203-89856203 on Build GRCh38
rs901299828 Chr.16: 89856326-89856326 on Build GRCh38
rs140601253 Chr.16: 89856208-89856208 on Build GRCh38
rs777732250 Chr.16: 89856191-89856191 on Build GRCh38
rs1007746702 Chr.16: 89856339-89856339 on Build GRCh38
rs918298208 Chr.16: 89856248-89856248 on Build GRCh38
rs774184541 Chr.16: 89856215-89856215 on Build GRCh38
rs919297303 Chr.16: 89856306-89856306 on Build GRCh38
rs138095625 Chr.16: 89856278-89856278 on Build GRCh38
rs775485672 Chr.16: 89856272-89856272 on Build GRCh38
rs746068875 Chr.16: 89856199-89856199 on Build GRCh38
rs570025429 Chr.16: 89856302-89856302 on Build GRCh38
rs539104710 Chr.16: 89856362-89856362 on Build GRCh38
rs199970599 Chr.16: 89856194-89856194 on Build GRCh38
rs750700313 Chr.16: 89856264-89856264 on Build GRCh38
rs201285166 Chr.16: 89856246-89856246 on Build GRCh38
rs28479168 Chr.16: 89856340-89856340 on Build GRCh38
rs890017433 Chr.16: 89856298-89856298 on Build GRCh38
rs371646113 Chr.16: 89856232-89856232 on Build GRCh38
rs758557329 Chr.16: 89856277-89856277 on Build GRCh38

View Full Product Details