Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65115868-65116129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TM7SF2
Entrez Gene ID
7108
Gene Name
transmembrane 7 superfamily member 2
Gene Aliases
ANG1, DHCR14A, NET47
UniGene
Hs.31130
Gene Chromosome Location
Chr.11: 65111854-65116235 on Build GRCh38
Transcripts
NM_001277233.1 NM_003273.3 NR_102367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5645957 c.1140C>T p.T380T Chr.11: 65115936-65115936 on Build GRCh38
6277486 c.1248C>T p.Y416Y Chr.11: 65116044-65116044 on Build GRCh38
3788024 c.1117T>C p.Y373H Chr.11: 65115913-65115913 on Build GRCh38
930329 c.1155C>T p.H385H Chr.11: 65115951-65115951 on Build GRCh38
5510517 c.1141G>A p.A381T Chr.11: 65115937-65115937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368355832 Chr.11: 65116053-65116053 on Build GRCh38
rs377534768 Chr.11: 65116019-65116019 on Build GRCh38
rs369161377 Chr.11: 65115922-65115922 on Build GRCh38
rs777690598 Chr.11: 65116078-65116078 on Build GRCh38
rs769961279 Chr.11: 65115953-65115953 on Build GRCh38
rs574235274 Chr.11: 65115962-65115962 on Build GRCh38
rs776795728 Chr.11: 65115949-65115949 on Build GRCh38
rs749953666 Chr.11: 65115892-65115892 on Build GRCh38
rs1013202329 Chr.11: 65116008-65116008 on Build GRCh38
rs770988729 Chr.11: 65116080-65116080 on Build GRCh38
rs370647941 Chr.11: 65116064-65116064 on Build GRCh38
rs760211079 Chr.11: 65115970-65115970 on Build GRCh38
rs111853188 Chr.11: 65115891-65115891 on Build GRCh38
rs771043618 Chr.11: 65116044-65116044 on Build GRCh38
rs372966759 Chr.11: 65115961-65115961 on Build GRCh38
rs912078312 Chr.11: 65116023-65116023 on Build GRCh38
rs780139717 Chr.11: 65116016-65116016 on Build GRCh38
rs757798946 Chr.11: 65116001-65116001 on Build GRCh38
rs750446909 Chr.11: 65116068-65116068 on Build GRCh38
rs903662919 Chr.11: 65115960-65115960 on Build GRCh38
rs895611462 Chr.11: 65115939-65115939 on Build GRCh38
rs756165697 Chr.11: 65116072-65116072 on Build GRCh38
rs764855560 Chr.11: 65115986-65115986 on Build GRCh38
rs375604515 Chr.11: 65116052-65116052 on Build GRCh38
rs1046912 Chr.11: 65116075-65116075 on Build GRCh38
rs754635636 Chr.11: 65115984-65115984 on Build GRCh38
rs370698238 Chr.11: 65116031-65116031 on Build GRCh38
rs1014191221 Chr.11: 65115940-65115940 on Build GRCh38
rs771022046 Chr.11: 65115946-65115946 on Build GRCh38
rs546739753 Chr.11: 65116058-65116058 on Build GRCh38
rs763791321 Chr.11: 65116102-65116102 on Build GRCh38
rs576235129 Chr.11: 65116017-65116017 on Build GRCh38
rs762488394 Chr.11: 65116101-65116101 on Build GRCh38
rs747033645 Chr.11: 65115944-65115944 on Build GRCh38
rs771788944 Chr.11: 65116024-65116024 on Build GRCh38
rs774186843 Chr.11: 65115954-65115954 on Build GRCh38
rs1023269696 Chr.11: 65115941-65115941 on Build GRCh38
rs112104203 Chr.11: 65115890-65115890 on Build GRCh38
rs748508503 Chr.11: 65115927-65115927 on Build GRCh38
rs17855608 Chr.11: 65116022-65116022 on Build GRCh38
rs770048261 Chr.11: 65116015-65116015 on Build GRCh38
rs975032673 Chr.11: 65116040-65116040 on Build GRCh38
rs752313771 Chr.11: 65116056-65116056 on Build GRCh38
rs753436919 Chr.11: 65115977-65115977 on Build GRCh38
rs764876825 Chr.11: 65116055-65116055 on Build GRCh38
rs769217632 Chr.11: 65116095-65116095 on Build GRCh38
rs746855481 Chr.11: 65116037-65116037 on Build GRCh38
rs766016020 Chr.11: 65115972-65115972 on Build GRCh38
rs544507657 Chr.11: 65115903-65115903 on Build GRCh38
rs781223700 Chr.11: 65116003-65116003 on Build GRCh38
rs746060735 Chr.11: 65115952-65115952 on Build GRCh38
rs367578927 Chr.11: 65116079-65116079 on Build GRCh38
rs759037503 Chr.11: 65116094-65116094 on Build GRCh38
rs371857068 Chr.11: 65116097-65116097 on Build GRCh38
rs760660829 Chr.11: 65116041-65116041 on Build GRCh38
rs1022794064 Chr.11: 65116111-65116111 on Build GRCh38
rs776714979 Chr.11: 65116082-65116082 on Build GRCh38
rs764409119 Chr.11: 65116061-65116061 on Build GRCh38
rs150748205 Chr.11: 65116073-65116073 on Build GRCh38
rs1012043850 Chr.11: 65115893-65115893 on Build GRCh38
rs755351709 Chr.11: 65115910-65115910 on Build GRCh38
rs1023683629 Chr.11: 65116036-65116036 on Build GRCh38
rs111613375 Chr.11: 65116112-65116112 on Build GRCh38
rs753563762 Chr.11: 65115909-65115909 on Build GRCh38
rs200277720 Chr.11: 65115965-65115965 on Build GRCh38
rs183160005 Chr.11: 65115990-65115990 on Build GRCh38
rs375239318 Chr.11: 65115895-65115895 on Build GRCh38
rs758924190 Chr.11: 65115936-65115936 on Build GRCh38
rs751966937 Chr.11: 65116063-65116063 on Build GRCh38
rs373299244 Chr.11: 65116009-65116009 on Build GRCh38
rs778294179 Chr.11: 65115991-65115991 on Build GRCh38
rs141628241 Chr.11: 65115996-65115996 on Build GRCh38
rs769069841 Chr.11: 65116075-65116075 on Build GRCh38
rs778148061 Chr.11: 65115938-65115938 on Build GRCh38

View Full Product Details