Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 87332282-87332555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASA1
Entrez Gene ID
5921
Gene Name
RAS p21 protein activator 1
Gene Aliases
CM-AVM, CMAVM, GAP, PKWS, RASA, RASGAP, p120, p120GAP, p120RASGAP
UniGene
Hs.664080
Gene Chromosome Location
Chr.5: 87267801-87391926 on Build GRCh38
Transcripts
NM_002890.2 NM_022650.2 XM_011543525.1 XM_011543527.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1695954 c.203G>A p.R68H Chr.5: 87332548-87332548 on Build GRCh38
6952876 c.710G>C p.G237A Chr.5: 87332524-87332524 on Build GRCh38
6948809 c.178G>T p.G60* Chr.5: 87332523-87332523 on Build GRCh38
6952877 c.179G>C p.G60A Chr.5: 87332524-87332524 on Build GRCh38
1695953 c.734G>A p.R245H Chr.5: 87332548-87332548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776195697 Chr.5: 87332487-87332488 on Build GRCh38
rs74492894 Chr.5: 87332349-87332349 on Build GRCh38
rs527807495 Chr.5: 87332496-87332496 on Build GRCh38
rs201997632 Chr.5: 87332508-87332508 on Build GRCh38
rs768103774 Chr.5: 87332473-87332473 on Build GRCh38
rs983046029 Chr.5: 87332389-87332389 on Build GRCh38
rs776898751 Chr.5: 87332466-87332466 on Build GRCh38
rs116288928 Chr.5: 87332392-87332392 on Build GRCh38
rs554871804 Chr.5: 87332341-87332341 on Build GRCh38
rs578169577 Chr.5: 87332357-87332357 on Build GRCh38
rs760074054 Chr.5: 87332483-87332483 on Build GRCh38
rs141147099 Chr.5: 87332316-87332316 on Build GRCh38
rs960236167 Chr.5: 87332413-87332413 on Build GRCh38
rs746630237 Chr.5: 87332472-87332472 on Build GRCh38
rs144964372 Chr.5: 87332374-87332374 on Build GRCh38
rs918751235 Chr.5: 87332410-87332410 on Build GRCh38
rs771321253 Chr.5: 87332456-87332456 on Build GRCh38
rs992950713 Chr.5: 87332487-87332487 on Build GRCh38
rs972814310 Chr.5: 87332408-87332408 on Build GRCh38
rs765878106 Chr.5: 87332485-87332485 on Build GRCh38
rs762566440 Chr.5: 87332525-87332525 on Build GRCh38

View Full Product Details