Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207050803-207051050 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YOD1
Entrez Gene ID
55432
Gene Name
YOD1 deubiquitinase
Gene Aliases
DUBA8, OTUD2, PRO0907
UniGene
Hs.567533
Gene Chromosome Location
Chr.1: 207043849-207052980 on Build GRCh38
Transcripts
NM_001276320.1 NM_018566.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5053028 c.120C>T p.G40G Chr.1: 207050911-207050911 on Build GRCh38
95550 c.197G>T p.R66L Chr.1: 207050834-207050834 on Build GRCh38
116566 c.158C>G p.A53G Chr.1: 207050873-207050873 on Build GRCh38
7398127 c.79G>A p.A27T Chr.1: 207050952-207050952 on Build GRCh38
4621575 c.54C>A p.F18L Chr.1: 207050977-207050977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776024148 Chr.1: 207050918-207050918 on Build GRCh38
rs906407342 Chr.1: 207051018-207051018 on Build GRCh38
rs765860156 Chr.1: 207050987-207050987 on Build GRCh38
rs78043860 Chr.1: 207050983-207050983 on Build GRCh38
rs764493387 Chr.1: 207050924-207050924 on Build GRCh38
rs541136434 Chr.1: 207050952-207050952 on Build GRCh38
rs900736502 Chr.1: 207050974-207050974 on Build GRCh38
rs545732408 Chr.1: 207050925-207050925 on Build GRCh38
rs761550399 Chr.1: 207050926-207050926 on Build GRCh38
rs1003314898 Chr.1: 207051026-207051026 on Build GRCh38
rs750013764 Chr.1: 207050823-207050823 on Build GRCh38
rs959425750 Chr.1: 207050944-207050944 on Build GRCh38
rs970532602 Chr.1: 207050840-207050840 on Build GRCh38
rs746107083 Chr.1: 207050957-207050957 on Build GRCh38
rs1039331999 Chr.1: 207050967-207050967 on Build GRCh38
rs756347464 Chr.1: 207051028-207051028 on Build GRCh38
rs983619989 Chr.1: 207050958-207050958 on Build GRCh38
rs112723983 Chr.1: 207050979-207050979 on Build GRCh38
rs116505937 Chr.1: 207050934-207050934 on Build GRCh38
rs534842552 Chr.1: 207051020-207051020 on Build GRCh38
rs754732226 Chr.1: 207050856-207050856 on Build GRCh38
rs753406894 Chr.1: 207050990-207050990 on Build GRCh38
rs749209354 Chr.1: 207050855-207050855 on Build GRCh38
rs757828294 Chr.1: 207050962-207050962 on Build GRCh38
rs532257170 Chr.1: 207050981-207050981 on Build GRCh38
rs770180166 Chr.1: 207050916-207050916 on Build GRCh38
rs767315302 Chr.1: 207050933-207050933 on Build GRCh38
rs780009774 Chr.1: 207050961-207050961 on Build GRCh38
rs769845525 Chr.1: 207050959-207050959 on Build GRCh38
rs576800385 Chr.1: 207050882-207050882 on Build GRCh38
rs747180418 Chr.1: 207050866-207050866 on Build GRCh38
rs200758571 Chr.1: 207050949-207050949 on Build GRCh38
rs561023930 Chr.1: 207050954-207050954 on Build GRCh38
rs530167183 Chr.1: 207050975-207050975 on Build GRCh38
rs565704574 Chr.1: 207050991-207050991 on Build GRCh38
rs778669007 Chr.1: 207050950-207050950 on Build GRCh38
rs777433140 Chr.1: 207051014-207051014 on Build GRCh38
rs776571331 Chr.1: 207050906-207050906 on Build GRCh38
rs1009770682 Chr.1: 207050994-207050994 on Build GRCh38
rs755597834 Chr.1: 207050938-207050938 on Build GRCh38
rs755970691 Chr.1: 207050830-207050830 on Build GRCh38
rs112604654 Chr.1: 207050980-207050980 on Build GRCh38
rs746000674 Chr.1: 207050907-207050907 on Build GRCh38
rs549752987 Chr.1: 207050976-207050976 on Build GRCh38
rs778687294 Chr.1: 207050865-207050865 on Build GRCh38
rs780626556 Chr.1: 207050883-207050883 on Build GRCh38
rs1033576762 Chr.1: 207051030-207051030 on Build GRCh38
rs758140321 Chr.1: 207051007-207051007 on Build GRCh38
rs753746933 Chr.1: 207050946-207050946 on Build GRCh38
rs201269070 Chr.1: 207050945-207050945 on Build GRCh38
rs563387440 Chr.1: 207050977-207050977 on Build GRCh38
rs765201156 Chr.1: 207050992-207050992 on Build GRCh38
rs763559031 Chr.1: 207050919-207050919 on Build GRCh38
rs775408983 Chr.1: 207050998-207050998 on Build GRCh38
rs780085839 Chr.1: 207050846-207050846 on Build GRCh38

View Full Product Details