Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 44624710-44624935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YIPF7
Entrez Gene ID
285525
Gene Name
Yip1 domain family member 7
Gene Aliases
FinGER9
UniGene
Hs.596000
Gene Chromosome Location
Chr.4: 44619578-44656868 on Build GRCh38
Transcripts
NM_182592.2 XM_011513679.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
447887 c.542T>A p.I181N Chr.4: 44624739-44624739 on Build GRCh38
5694291 c.533T>A p.M178K Chr.4: 44624748-44624748 on Build GRCh38
5697947 c.502G>T p.G168* Chr.4: 44624779-44624779 on Build GRCh38
6198843 c.531C>A p.G177G Chr.4: 44624750-44624750 on Build GRCh38
4941184 c.564C>T p.A188A Chr.4: 44624717-44624717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902140642 Chr.4: 44624842-44624842 on Build GRCh38
rs946435384 Chr.4: 44624825-44624825 on Build GRCh38
rs368914025 Chr.4: 44624768-44624768 on Build GRCh38
rs763233871 Chr.4: 44624744-44624744 on Build GRCh38
rs1033028619 Chr.4: 44624865-44624865 on Build GRCh38
rs148043580 Chr.4: 44624896-44624896 on Build GRCh38
rs746483740 Chr.4: 44624823-44624823 on Build GRCh38
rs764265212 Chr.4: 44624747-44624747 on Build GRCh38
rs746359605 Chr.4: 44624828-44624828 on Build GRCh38
rs984246706 Chr.4: 44624783-44624783 on Build GRCh38
rs756724997 Chr.4: 44624824-44624824 on Build GRCh38
rs957834590 Chr.4: 44624890-44624890 on Build GRCh38
rs771017123 Chr.4: 44624754-44624755 on Build GRCh38
rs758043645 Chr.4: 44624790-44624790 on Build GRCh38
rs759124439 Chr.4: 44624813-44624814 on Build GRCh38
rs767080266 Chr.4: 44624822-44624822 on Build GRCh38
rs17600797 Chr.4: 44624789-44624789 on Build GRCh38
rs542917133 Chr.4: 44624843-44624843 on Build GRCh38
rs6838327 Chr.4: 44624829-44624829 on Build GRCh38
rs566350509 Chr.4: 44624770-44624770 on Build GRCh38
rs1008095955 Chr.4: 44624792-44624792 on Build GRCh38
rs753834911 Chr.4: 44624773-44624773 on Build GRCh38
rs774468579 Chr.4: 44624749-44624749 on Build GRCh38
rs775888663 Chr.4: 44624742-44624742 on Build GRCh38
rs777309760 Chr.4: 44624793-44624793 on Build GRCh38
rs764598281 Chr.4: 44624763-44624763 on Build GRCh38
rs939624291 Chr.4: 44624817-44624817 on Build GRCh38
rs781620640 Chr.4: 44624826-44624826 on Build GRCh38
rs996080602 Chr.4: 44624911-44624911 on Build GRCh38
rs770155368 Chr.4: 44624732-44624732 on Build GRCh38
rs928204944 Chr.4: 44624815-44624815 on Build GRCh38
rs754765822 Chr.4: 44624781-44624781 on Build GRCh38
rs368044160 Chr.4: 44624800-44624800 on Build GRCh38
rs190942228 Chr.4: 44624820-44624820 on Build GRCh38
rs901758382 Chr.4: 44624884-44624884 on Build GRCh38
rs369845305 Chr.4: 44624873-44624873 on Build GRCh38
rs774391604 Chr.4: 44624769-44624770 on Build GRCh38

View Full Product Details