Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 21157957-21158230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKA3
Entrez Gene ID
221150
Gene Name
spindle and kinetochore associated complex subunit 3
Gene Aliases
C13orf3, RAMA1
UniGene
Hs.88523
Gene Chromosome Location
Chr.13: 21153595-21176602 on Build GRCh38
Transcripts
NM_001166017.1 NM_145061.5 XM_005266288.3 XM_011534994.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6338937 c.1073C>T p.T358I Chr.13: 21157968-21157968 on Build GRCh38
7155796 c.1049C>T p.S350F Chr.13: 21157992-21157992 on Build GRCh38
7155797 c.1049C>T p.S350F Chr.13: 21157992-21157992 on Build GRCh38
6284813 c.1031C>T p.P344L Chr.13: 21158010-21158010 on Build GRCh38
3987329 c.984T>C p.T328T Chr.13: 21158057-21158057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781775512 Chr.13: 21158132-21158132 on Build GRCh38
rs17279819 Chr.13: 21158036-21158036 on Build GRCh38
rs759208587 Chr.13: 21158024-21158024 on Build GRCh38
rs781167794 Chr.13: 21158140-21158141 on Build GRCh38
rs376831484 Chr.13: 21158062-21158062 on Build GRCh38
rs777447983 Chr.13: 21158162-21158162 on Build GRCh38
rs941597275 Chr.13: 21158033-21158033 on Build GRCh38
rs752414983 Chr.13: 21158113-21158113 on Build GRCh38
rs777372016 Chr.13: 21158162-21158164 on Build GRCh38
rs750946749 Chr.13: 21158134-21158134 on Build GRCh38
rs768861452 Chr.13: 21158160-21158160 on Build GRCh38
rs761742758 Chr.13: 21157993-21157993 on Build GRCh38
rs147108341 Chr.13: 21158077-21158077 on Build GRCh38
rs757420168 Chr.13: 21158132-21158133 on Build GRCh38
rs764380078 Chr.13: 21158125-21158125 on Build GRCh38
rs765383514 Chr.13: 21158132-21158134 on Build GRCh38
rs770849089 Chr.13: 21158093-21158093 on Build GRCh38
rs748140660 Chr.13: 21158147-21158155 on Build GRCh38
rs772306054 Chr.13: 21158199-21158199 on Build GRCh38
rs754879051 Chr.13: 21158125-21158126 on Build GRCh38
rs756156277 Chr.13: 21158158-21158158 on Build GRCh38
rs754787862 Chr.13: 21158137-21158138 on Build GRCh38
rs760654135 Chr.13: 21158008-21158008 on Build GRCh38
rs181879406 Chr.13: 21157991-21157991 on Build GRCh38
rs769967583 Chr.13: 21158064-21158064 on Build GRCh38
rs776597503 Chr.13: 21158171-21158171 on Build GRCh38
rs775916015 Chr.13: 21158129-21158130 on Build GRCh38
rs748823604 Chr.13: 21158162-21158166 on Build GRCh38
rs746266876 Chr.13: 21158065-21158065 on Build GRCh38
rs776707291 Chr.13: 21157979-21157979 on Build GRCh38
rs865785583 Chr.13: 21157986-21157986 on Build GRCh38
rs748913598 Chr.13: 21158128-21158129 on Build GRCh38
rs778963799 Chr.13: 21158055-21158055 on Build GRCh38
rs771827485 Chr.13: 21158080-21158080 on Build GRCh38
rs749473787 Chr.13: 21158155-21158155 on Build GRCh38
rs138407947 Chr.13: 21158116-21158116 on Build GRCh38
rs201402604 Chr.13: 21157997-21157997 on Build GRCh38
rs770599511 Chr.13: 21158168-21158168 on Build GRCh38
rs756322128 Chr.13: 21158136-21158136 on Build GRCh38
rs772989846 Chr.13: 21158083-21158083 on Build GRCh38
rs751367657 Chr.13: 21158135-21158135 on Build GRCh38
rs1056394914 Chr.13: 21158148-21158148 on Build GRCh38
rs148901267 Chr.13: 21157998-21157998 on Build GRCh38
rs751903665 Chr.13: 21158127-21158127 on Build GRCh38
rs781154501 Chr.13: 21158061-21158061 on Build GRCh38
rs753283392 Chr.13: 21158016-21158016 on Build GRCh38
rs4111077 Chr.13: 21158209-21158209 on Build GRCh38
rs776150162 Chr.13: 21158095-21158095 on Build GRCh38
rs770095678 Chr.13: 21157985-21157985 on Build GRCh38
rs760718319 Chr.13: 21158130-21158131 on Build GRCh38
rs753948268 Chr.13: 21158131-21158132 on Build GRCh38
rs908834307 Chr.13: 21158013-21158013 on Build GRCh38
rs17345571 Chr.13: 21158054-21158054 on Build GRCh38
rs200780567 Chr.13: 21158011-21158011 on Build GRCh38
rs759086059 Chr.13: 21158101-21158101 on Build GRCh38
rs780274906 Chr.13: 21158142-21158142 on Build GRCh38
rs760538954 Chr.13: 21158086-21158086 on Build GRCh38
rs371829916 Chr.13: 21158163-21158163 on Build GRCh38
rs746101953 Chr.13: 21157984-21157984 on Build GRCh38
rs939231200 Chr.13: 21158154-21158154 on Build GRCh38
rs541061764 Chr.13: 21158129-21158129 on Build GRCh38
rs917802472 Chr.13: 21158166-21158166 on Build GRCh38
rs753244984 Chr.13: 21158126-21158127 on Build GRCh38
rs764702773 Chr.13: 21158111-21158111 on Build GRCh38

View Full Product Details