Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201359538-201359775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNNT2
Entrez Gene ID
7139
Gene Name
troponin T2, cardiac type
Gene Aliases
CMD1D, CMH2, CMPD2, LVNC6, RCM3, TnTC, cTnT
UniGene
Hs.533613
Gene Chromosome Location
Chr.1: 201359008-201377828 on Build GRCh38
Transcripts
NM_000364.3 NM_001001430.2 NM_001001431.2 NM_001001432.2 NM_001276345.1 NM_001276346.1 NM_001276347.1 XM_006711508.3 XM_006711509.3 XM_011509938.2 XM_011509939.1 XM_011509940.2 XM_011509941.2 XM_011509942.2 XM_011509943.2 XM_011509944.2 XM_011509946.1 XM_017002216.1 XM_017002217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4863831 c.779G>A p.R260Q Chr.1: 201359650-201359650 on Build GRCh38
5511628 c.834C>T p.N278N Chr.1: 201359637-201359637 on Build GRCh38
902175 c.785G>A p.R262Q Chr.1: 201359650-201359650 on Build GRCh38
5511627 c.798C>T p.N266N Chr.1: 201359637-201359637 on Build GRCh38
5511629 c.792C>T p.N264N Chr.1: 201359637-201359637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs730881119 Chr.1: 201359629-201359630 on Build GRCh38
rs756065564 Chr.1: 201359705-201359705 on Build GRCh38
rs1005243006 Chr.1: 201359596-201359596 on Build GRCh38
rs376739754 Chr.1: 201359579-201359579 on Build GRCh38
rs786204408 Chr.1: 201359649-201359649 on Build GRCh38
rs755148270 Chr.1: 201359592-201359592 on Build GRCh38
rs2275863 Chr.1: 201359696-201359696 on Build GRCh38
rs990771026 Chr.1: 201359647-201359647 on Build GRCh38
rs796925245 Chr.1: 201359630-201359630 on Build GRCh38
rs1021018020 Chr.1: 201359733-201359733 on Build GRCh38
rs768470426 Chr.1: 201359658-201359658 on Build GRCh38
rs12043978 Chr.1: 201359749-201359749 on Build GRCh38
rs374622439 Chr.1: 201359586-201359586 on Build GRCh38
rs376923877 Chr.1: 201359637-201359637 on Build GRCh38
rs942218410 Chr.1: 201359737-201359737 on Build GRCh38
rs796311418 Chr.1: 201359622-201359623 on Build GRCh38
rs397516483 Chr.1: 201359659-201359659 on Build GRCh38
rs754253675 Chr.1: 201359684-201359684 on Build GRCh38
rs45495692 Chr.1: 201359601-201359601 on Build GRCh38
rs759864879 Chr.1: 201359690-201359690 on Build GRCh38
rs768313123 Chr.1: 201359573-201359573 on Build GRCh38
rs748970759 Chr.1: 201359651-201359651 on Build GRCh38
rs202146260 Chr.1: 201359600-201359600 on Build GRCh38
rs575803809 Chr.1: 201359590-201359590 on Build GRCh38
rs1038489479 Chr.1: 201359744-201359744 on Build GRCh38
rs111377893 Chr.1: 201359622-201359622 on Build GRCh38
rs368131332 Chr.1: 201359699-201359699 on Build GRCh38
rs730881125 Chr.1: 201359650-201359650 on Build GRCh38
rs371552542 Chr.1: 201359713-201359713 on Build GRCh38
rs761288511 Chr.1: 201359661-201359661 on Build GRCh38
rs121964861 Chr.1: 201359636-201359636 on Build GRCh38
rs113020380 Chr.1: 201359620-201359620 on Build GRCh38
rs863225119 Chr.1: 201359632-201359632 on Build GRCh38
rs191930037 Chr.1: 201359585-201359585 on Build GRCh38
rs761131892 Chr.1: 201359678-201359678 on Build GRCh38
rs1002828059 Chr.1: 201359680-201359680 on Build GRCh38
rs371384395 Chr.1: 201359662-201359662 on Build GRCh38
rs765447758 Chr.1: 201359691-201359691 on Build GRCh38
rs4523540 Chr.1: 201359639-201359639 on Build GRCh38
rs917942614 Chr.1: 201359676-201359676 on Build GRCh38
rs193922620 Chr.1: 201359618-201359618 on Build GRCh38
rs758497752 Chr.1: 201359606-201359606 on Build GRCh38
rs766756410 Chr.1: 201359681-201359681 on Build GRCh38
rs780117607 Chr.1: 201359711-201359711 on Build GRCh38
rs773596960 Chr.1: 201359668-201359668 on Build GRCh38
rs752947789 Chr.1: 201359695-201359695 on Build GRCh38
rs45576635 Chr.1: 201359577-201359577 on Build GRCh38
rs45570832 Chr.1: 201359580-201359580 on Build GRCh38

View Full Product Details