Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37084354-37084611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMPRSS6
Entrez Gene ID
164656
Gene Name
transmembrane protease, serine 6
Gene Aliases
IRIDA
UniGene
Hs.370885
Gene Chromosome Location
Chr.22: 37065436-37109701 on Build GRCh38
Transcripts
NM_001289000.1 NM_001289001.1 NM_153609.3 XM_011529989.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3150132 c.1121C>G p.S374C Chr.22: 37084397-37084397 on Build GRCh38
3150131 c.1121C>G p.S374C Chr.22: 37084397-37084397 on Build GRCh38
4103891 c.1163T>C p.L388P Chr.22: 37084355-37084355 on Build GRCh38
5423991 c.1132G>A p.G378S Chr.22: 37084386-37084386 on Build GRCh38
5936743 c.1087-5C>T p.? Chr.22: 37084409-37084409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567122650 Chr.22: 37084423-37084423 on Build GRCh38
rs73160055 Chr.22: 37084565-37084565 on Build GRCh38
rs759588829 Chr.22: 37084381-37084381 on Build GRCh38
rs762853689 Chr.22: 37084434-37084460 on Build GRCh38
rs368625259 Chr.22: 37084419-37084419 on Build GRCh38
rs992528088 Chr.22: 37084462-37084462 on Build GRCh38
rs1022762506 Chr.22: 37084392-37084392 on Build GRCh38
rs769837217 Chr.22: 37084415-37084415 on Build GRCh38
rs368672399 Chr.22: 37084377-37084377 on Build GRCh38
rs534839740 Chr.22: 37084553-37084554 on Build GRCh38
rs765448423 Chr.22: 37084437-37084437 on Build GRCh38
rs775257747 Chr.22: 37084426-37084436 on Build GRCh38
rs763126870 Chr.22: 37084450-37084450 on Build GRCh38
rs777790042 Chr.22: 37084395-37084395 on Build GRCh38
rs1050079094 Chr.22: 37084487-37084487 on Build GRCh38
rs900968153 Chr.22: 37084442-37084442 on Build GRCh38
rs145585962 Chr.22: 37084417-37084417 on Build GRCh38
rs751272393 Chr.22: 37084401-37084401 on Build GRCh38
rs757116898 Chr.22: 37084402-37084402 on Build GRCh38
rs371438621 Chr.22: 37084427-37084427 on Build GRCh38
rs760913680 Chr.22: 37084420-37084420 on Build GRCh38
rs371733127 Chr.22: 37084446-37084446 on Build GRCh38
rs768896425 Chr.22: 37084387-37084387 on Build GRCh38
rs777158187 Chr.22: 37084560-37084560 on Build GRCh38
rs893711031 Chr.22: 37084492-37084492 on Build GRCh38
rs546400688 Chr.22: 37084509-37084509 on Build GRCh38
rs1023987008 Chr.22: 37084533-37084533 on Build GRCh38
rs375628093 Chr.22: 37084418-37084418 on Build GRCh38
rs368410991 Chr.22: 37084440-37084440 on Build GRCh38
rs1020909864 Chr.22: 37084567-37084567 on Build GRCh38
rs745578563 Chr.22: 37084416-37084416 on Build GRCh38
rs528314053 Chr.22: 37084499-37084499 on Build GRCh38
rs1004004592 Chr.22: 37084589-37084589 on Build GRCh38
rs765356650 Chr.22: 37084384-37084384 on Build GRCh38
rs780690433 Chr.22: 37084413-37084413 on Build GRCh38
rs188110925 Chr.22: 37084569-37084569 on Build GRCh38
rs762548397 Chr.22: 37084386-37084386 on Build GRCh38

View Full Product Details