Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75690272-75690501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POC5
Entrez Gene ID
134359
Gene Name
POC5 centriolar protein
Gene Aliases
C5orf37
UniGene
Hs.432726
Gene Chromosome Location
Chr.5: 75674198-75717488 on Build GRCh38
Transcripts
NM_001099271.1 NM_152408.2 XM_005248436.1 XM_011543158.2 XM_011543159.1 XM_011543160.1 XM_017009037.1 XM_017009038.1 XM_017009039.1 XM_017009040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
739228 c.867G>C p.Q289H Chr.5: 75690416-75690416 on Build GRCh38
739227 c.942G>C p.Q314H Chr.5: 75690416-75690416 on Build GRCh38
5508043 c.975G>A p.M325I Chr.5: 75690383-75690383 on Build GRCh38
3073718 c.796G>A p.V266I Chr.5: 75690487-75690487 on Build GRCh38
3073717 c.871G>A p.V291I Chr.5: 75690487-75690487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370019616 Chr.5: 75690409-75690409 on Build GRCh38
rs758466399 Chr.5: 75690440-75690440 on Build GRCh38
rs776233538 Chr.5: 75690344-75690344 on Build GRCh38
rs745663817 Chr.5: 75690347-75690347 on Build GRCh38
rs202066997 Chr.5: 75690453-75690453 on Build GRCh38
rs752837507 Chr.5: 75690431-75690431 on Build GRCh38
rs768078413 Chr.5: 75690397-75690397 on Build GRCh38
rs899482207 Chr.5: 75690447-75690447 on Build GRCh38
rs916293563 Chr.5: 75690402-75690402 on Build GRCh38
rs769546346 Chr.5: 75690351-75690351 on Build GRCh38
rs763650948 Chr.5: 75690378-75690378 on Build GRCh38
rs865950781 Chr.5: 75690327-75690327 on Build GRCh38
rs201228814 Chr.5: 75690377-75690377 on Build GRCh38
rs773434738 Chr.5: 75690375-75690375 on Build GRCh38
rs1007808108 Chr.5: 75690412-75690412 on Build GRCh38
rs377357203 Chr.5: 75690472-75690472 on Build GRCh38
rs751857490 Chr.5: 75690383-75690383 on Build GRCh38
rs745837144 Chr.5: 75690470-75690470 on Build GRCh38
rs754034926 Chr.5: 75690414-75690414 on Build GRCh38
rs778898151 Chr.5: 75690423-75690423 on Build GRCh38
rs1046035278 Chr.5: 75690436-75690436 on Build GRCh38
rs766769077 Chr.5: 75690411-75690411 on Build GRCh38
rs377211158 Chr.5: 75690342-75690342 on Build GRCh38
rs762745790 Chr.5: 75690368-75690368 on Build GRCh38
rs755213651 Chr.5: 75690421-75690421 on Build GRCh38
rs775256136 Chr.5: 75690365-75690365 on Build GRCh38
rs762157133 Chr.5: 75690394-75690394 on Build GRCh38

View Full Product Details