Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57135570-57135795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZIM3
Entrez Gene ID
114026
Gene Name
zinc finger imprinted 3
Gene Aliases
ZNF657
UniGene
Hs.348622
Gene Chromosome Location
Chr.19: 57134096-57145202 on Build GRCh38
Transcripts
NM_052882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4555532 c.658G>A p.E220K Chr.19: 57135679-57135679 on Build GRCh38
7115530 c.547C>G p.Q183E Chr.19: 57135790-57135790 on Build GRCh38
3540342 c.553C>T p.H185Y Chr.19: 57135784-57135784 on Build GRCh38
5464785 c.671A>C p.K224T Chr.19: 57135666-57135666 on Build GRCh38
3540340 c.686G>A p.G229E Chr.19: 57135651-57135651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761009741 Chr.19: 57135655-57135655 on Build GRCh38
rs1031472257 Chr.19: 57135744-57135744 on Build GRCh38
rs35202951 Chr.19: 57135724-57135724 on Build GRCh38
rs754252389 Chr.19: 57135673-57135673 on Build GRCh38
rs773879897 Chr.19: 57135602-57135602 on Build GRCh38
rs764954740 Chr.19: 57135770-57135770 on Build GRCh38
rs755002491 Chr.19: 57135700-57135701 on Build GRCh38
rs760090714 Chr.19: 57135723-57135723 on Build GRCh38
rs766656949 Chr.19: 57135626-57135626 on Build GRCh38
rs773826974 Chr.19: 57135703-57135703 on Build GRCh38
rs777724086 Chr.19: 57135733-57135733 on Build GRCh38
rs754187075 Chr.19: 57135721-57135721 on Build GRCh38
rs531319362 Chr.19: 57135685-57135685 on Build GRCh38
rs200666945 Chr.19: 57135709-57135709 on Build GRCh38
rs374275027 Chr.19: 57135743-57135743 on Build GRCh38
rs766718644 Chr.19: 57135715-57135715 on Build GRCh38
rs149938645 Chr.19: 57135738-57135738 on Build GRCh38
rs867628159 Chr.19: 57135679-57135679 on Build GRCh38
rs761620269 Chr.19: 57135742-57135742 on Build GRCh38
rs930978385 Chr.19: 57135619-57135622 on Build GRCh38
rs761369261 Chr.19: 57135704-57135704 on Build GRCh38
rs150862431 Chr.19: 57135688-57135688 on Build GRCh38
rs779864336 Chr.19: 57135768-57135768 on Build GRCh38
rs938763059 Chr.19: 57135625-57135625 on Build GRCh38
rs201068723 Chr.19: 57135713-57135713 on Build GRCh38
rs896790860 Chr.19: 57135601-57135601 on Build GRCh38
rs367996028 Chr.19: 57135686-57135686 on Build GRCh38
rs143025400 Chr.19: 57135607-57135607 on Build GRCh38
rs34471789 Chr.19: 57135674-57135674 on Build GRCh38
rs146062644 Chr.19: 57135622-57135622 on Build GRCh38
rs768617497 Chr.19: 57135699-57135699 on Build GRCh38
rs878866223 Chr.19: 57135706-57135706 on Build GRCh38
rs750738318 Chr.19: 57135617-57135617 on Build GRCh38
rs370291347 Chr.19: 57135739-57135739 on Build GRCh38
rs368695053 Chr.19: 57135687-57135687 on Build GRCh38
rs758622269 Chr.19: 57135725-57135725 on Build GRCh38
rs767205997 Chr.19: 57135613-57135613 on Build GRCh38
rs7251328 Chr.19: 57135647-57135647 on Build GRCh38
rs754315030 Chr.19: 57135637-57135637 on Build GRCh38

View Full Product Details