Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 25604715-25604956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOP2B
Entrez Gene ID
7155
Gene Name
topoisomerase (DNA) II beta
Gene Aliases
TOPIIB, top2beta
UniGene
Hs.475733
Gene Chromosome Location
Chr.3: 25597905-25664936 on Build GRCh38
Transcripts
NM_001068.3 XM_005265427.3 XM_011534057.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6806187 c.4466C>A p.A1489D Chr.3: 25604768-25604768 on Build GRCh38
3660465 c.4474+3A>G p.? Chr.3: 25604757-25604757 on Build GRCh38
6510949 c.4474G>T p.G1492* Chr.3: 25604760-25604760 on Build GRCh38
6356544 c.4462G>A p.A1488T Chr.3: 25604772-25604772 on Build GRCh38
264141 c.4393G>T p.D1465Y Chr.3: 25604841-25604841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771418956 Chr.3: 25604840-25604840 on Build GRCh38
rs758347808 Chr.3: 25604755-25604755 on Build GRCh38
rs763705195 Chr.3: 25604879-25604879 on Build GRCh38
rs774107881 Chr.3: 25604818-25604818 on Build GRCh38
rs994503531 Chr.3: 25604891-25604891 on Build GRCh38
rs943061738 Chr.3: 25604881-25604881 on Build GRCh38
rs565600644 Chr.3: 25604898-25604898 on Build GRCh38
rs749064753 Chr.3: 25604784-25604784 on Build GRCh38
rs374330944 Chr.3: 25604915-25604915 on Build GRCh38
rs745398662 Chr.3: 25604822-25604822 on Build GRCh38
rs750826123 Chr.3: 25604900-25604900 on Build GRCh38
rs551760024 Chr.3: 25604854-25604854 on Build GRCh38
rs776193919 Chr.3: 25604904-25604904 on Build GRCh38
rs762113416 Chr.3: 25604894-25604894 on Build GRCh38
rs780325291 Chr.3: 25604762-25604762 on Build GRCh38
rs61751634 Chr.3: 25604777-25604777 on Build GRCh38
rs1022708055 Chr.3: 25604778-25604778 on Build GRCh38
rs747263680 Chr.3: 25604768-25604768 on Build GRCh38
rs954666241 Chr.3: 25604823-25604823 on Build GRCh38
rs370666167 Chr.3: 25604867-25604867 on Build GRCh38
rs755241213 Chr.3: 25604776-25604776 on Build GRCh38
rs988143045 Chr.3: 25604923-25604923 on Build GRCh38
rs895655528 Chr.3: 25604769-25604769 on Build GRCh38
rs771023574 Chr.3: 25604790-25604790 on Build GRCh38
rs758937149 Chr.3: 25604901-25604901 on Build GRCh38

View Full Product Details