Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 42387439-42387668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPHLN1
Entrez Gene ID
51535
Gene Name
periphilin 1
Gene Aliases
CR, HSPC206, HSPC232
UniGene
Hs.444157
Gene Chromosome Location
Chr.12: 42326126-42448621 on Build GRCh38
Transcripts
NM_001143787.1 NM_001143788.1 NM_001143789.1 NM_016488.6 NM_201438.1 NM_201439.1 NM_201440.1 NM_201515.1 XM_005268962.3 XM_011538459.2 XM_011538465.2 XM_011538467.2 XM_011538470.2 XM_011538471.2 XM_017019434.1 XM_017019435.1 XM_017019436.1 XM_017019437.1 XM_017019438.1 XM_017019439.1 XM_017019440.1 XM_017019441.1 XM_017019442.1 XM_017019443.1 XM_017019444.1 XM_017019445.1 XM_017019446.1 XM_017019447.1 XM_017019448.1 XM_017019449.1 XM_017019450.1 XM_017019451.1 XM_017019452.1 XM_017019453.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6753424 c.608A>T p.D203V Chr.12: 42387495-42387495 on Build GRCh38
6753422 c.443A>T p.D148V Chr.12: 42387495-42387495 on Build GRCh38
6258242 c.460G>T p.G154C Chr.12: 42387512-42387512 on Build GRCh38
3384344 c.644C>G p.S215* Chr.12: 42387531-42387531 on Build GRCh38
6258243 c.646G>T p.G216C Chr.12: 42387512-42387512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764138164 Chr.12: 42387586-42387586 on Build GRCh38
rs374534618 Chr.12: 42387485-42387485 on Build GRCh38
rs749793965 Chr.12: 42387502-42387502 on Build GRCh38
rs769247769 Chr.12: 42387503-42387503 on Build GRCh38
rs779125538 Chr.12: 42387508-42387508 on Build GRCh38
rs556118803 Chr.12: 42387518-42387518 on Build GRCh38
rs773500450 Chr.12: 42387524-42387524 on Build GRCh38
rs774173455 Chr.12: 42387587-42387587 on Build GRCh38
rs368593861 Chr.12: 42387574-42387574 on Build GRCh38
rs1015113520 Chr.12: 42387578-42387578 on Build GRCh38
rs937647657 Chr.12: 42387494-42387494 on Build GRCh38
rs771119579 Chr.12: 42387556-42387556 on Build GRCh38
rs141145173 Chr.12: 42387585-42387585 on Build GRCh38
rs776915694 Chr.12: 42387558-42387558 on Build GRCh38
rs755552152 Chr.12: 42387492-42387492 on Build GRCh38
rs779089561 Chr.12: 42387563-42387565 on Build GRCh38
rs779603127 Chr.12: 42387493-42387493 on Build GRCh38
rs747247433 Chr.12: 42387540-42387540 on Build GRCh38

View Full Product Details