Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43416891-43417138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102724428
Entrez Gene ID
102724428
Gene Name
serine/threonine-protein kinase SIK1
Gene Aliases
-
UniGene
Hs.282113
Gene Chromosome Location
Chr.21: 6111134-6123778 on Build GRCh38
Transcripts
NM_001320643.1 XM_017028208.1 NM_173354.3 XM_011529474.2

Gene Symbol
SIK1
Entrez Gene ID
150094
Gene Name
salt inducible kinase 1
Gene Aliases
EIEE30, MSK, SIK, SNF1LK
UniGene
Hs.282113
Gene Chromosome Location
Chr.21: 43414490-43427126 on Build GRCh38
Transcripts
NM_001320643.1 XM_017028208.1 NM_173354.3 XM_011529474.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4134967 c.2087C>T p.P696L Chr.21: 43417007-43417007 on Build GRCh38
6391596 c.2129C>T p.P710L Chr.21: 43416965-43416965 on Build GRCh38
4134968 c.2053G>T p.V685L Chr.21: 43417041-43417041 on Build GRCh38
1327383 c.2192A>T p.H731L Chr.21: 43416902-43416902 on Build GRCh38
4403353 c.2029C>T p.P677S Chr.21: 43417065-43417065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372469113 Chr.21: 43416919-43416919 on Build GRCh38
rs550744437 Chr.21: 43416976-43416976 on Build GRCh38
rs1049188125 Chr.21: 43416933-43416933 on Build GRCh38
rs534154863 Chr.21: 43417078-43417078 on Build GRCh38
rs778637849 Chr.21: 43417008-43417008 on Build GRCh38
rs776741814 Chr.21: 43416946-43416946 on Build GRCh38
rs779483942 Chr.21: 43417003-43417003 on Build GRCh38
rs926567754 Chr.21: 43416912-43416912 on Build GRCh38
rs766433765 Chr.21: 43416945-43416945 on Build GRCh38
rs754477690 Chr.21: 43417092-43417092 on Build GRCh38
rs772137462 Chr.21: 43416941-43416941 on Build GRCh38
rs763160252 Chr.21: 43417061-43417061 on Build GRCh38
rs553014547 Chr.21: 43417094-43417094 on Build GRCh38
rs35596465 Chr.21: 43416920-43416920 on Build GRCh38
rs367955174 Chr.21: 43417105-43417105 on Build GRCh38
rs547049916 Chr.21: 43416935-43416935 on Build GRCh38
rs778549988 Chr.21: 43417093-43417093 on Build GRCh38
rs892118949 Chr.21: 43417095-43417095 on Build GRCh38
rs777477090 Chr.21: 43417111-43417111 on Build GRCh38
rs567040219 Chr.21: 43417058-43417058 on Build GRCh38
rs775749558 Chr.21: 43417060-43417060 on Build GRCh38
rs560718043 Chr.21: 43416923-43416923 on Build GRCh38
rs56386767 Chr.21: 43417007-43417007 on Build GRCh38
rs759724871 Chr.21: 43416952-43416952 on Build GRCh38
rs929370860 Chr.21: 43417042-43417042 on Build GRCh38
rs926515568 Chr.21: 43417011-43417011 on Build GRCh38
rs200810453 Chr.21: 43417022-43417022 on Build GRCh38
rs1039205387 Chr.21: 43417062-43417062 on Build GRCh38
rs768025261 Chr.21: 43417082-43417082 on Build GRCh38
rs757755073 Chr.21: 43417001-43417001 on Build GRCh38
rs753064938 Chr.21: 43416968-43416968 on Build GRCh38
rs988303758 Chr.21: 43416974-43416974 on Build GRCh38
rs773124142 Chr.21: 43416982-43416982 on Build GRCh38
rs776595772 Chr.21: 43417046-43417046 on Build GRCh38
rs549619122 Chr.21: 43417032-43417032 on Build GRCh38
rs376915289 Chr.21: 43417006-43417006 on Build GRCh38
rs753288564 Chr.21: 43417089-43417089 on Build GRCh38
rs765405323 Chr.21: 43416960-43416960 on Build GRCh38
rs994009828 Chr.21: 43416928-43416928 on Build GRCh38
rs374537813 Chr.21: 43416942-43416942 on Build GRCh38
rs757730365 Chr.21: 43417081-43417081 on Build GRCh38
rs529008487 Chr.21: 43416940-43416940 on Build GRCh38
rs751934560 Chr.21: 43417080-43417080 on Build GRCh38
rs752544370 Chr.21: 43417035-43417035 on Build GRCh38
rs943521135 Chr.21: 43417057-43417057 on Build GRCh38
rs897883794 Chr.21: 43416922-43416922 on Build GRCh38
rs772047499 Chr.21: 43417029-43417029 on Build GRCh38
rs752076447 Chr.21: 43416985-43416985 on Build GRCh38
rs374313941 Chr.21: 43416973-43416973 on Build GRCh38
rs373872652 Chr.21: 43417045-43417045 on Build GRCh38
rs532529872 Chr.21: 43416943-43416943 on Build GRCh38
rs144982159 Chr.21: 43416936-43416936 on Build GRCh38
rs371001336 Chr.21: 43416934-43416934 on Build GRCh38

View Full Product Details