Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 55369935-55370186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRD5A3
Entrez Gene ID
79644
Gene Name
steroid 5 alpha-reductase 3
Gene Aliases
CDG1P, CDG1Q, KRIZI, SRD5A2L, SRD5A2L1
UniGene
Hs.39311
Gene Chromosome Location
Chr.4: 55346114-55373100 on Build GRCh38
Transcripts
NM_024592.4 XM_005265766.3 XM_005265767.3 XM_017008601.1 NR_037969.1

Gene Symbol
SRD5A3-AS1
Entrez Gene ID
100506462
Gene Name
SRD5A3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.718667
Gene Chromosome Location
Chr.4: 55366601-55385580 on Build GRCh38
Transcripts
NM_024592.4 XM_005265766.3 XM_005265767.3 XM_017008601.1 NR_037969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1243640 c.921G>A p.P307P Chr.4: 55370055-55370055 on Build GRCh38
1310144 c.882C>T p.L294L Chr.4: 55370016-55370016 on Build GRCh38
247596 c.914C>A p.S305Y Chr.4: 55370048-55370048 on Build GRCh38
1056157 c.890A>C p.Q297P Chr.4: 55370024-55370024 on Build GRCh38
4125195 c.942A>G p.L314L Chr.4: 55370076-55370076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201588517 Chr.4: 55370122-55370122 on Build GRCh38
rs778832270 Chr.4: 55369956-55369956 on Build GRCh38
rs565935886 Chr.4: 55370081-55370085 on Build GRCh38
rs769885824 Chr.4: 55370054-55370054 on Build GRCh38
rs756186617 Chr.4: 55370105-55370105 on Build GRCh38
rs770464192 Chr.4: 55370008-55370008 on Build GRCh38
rs764783481 Chr.4: 55370033-55370033 on Build GRCh38
rs769200099 Chr.4: 55370044-55370044 on Build GRCh38
rs747536860 Chr.4: 55370024-55370038 on Build GRCh38
rs763516132 Chr.4: 55370055-55370055 on Build GRCh38
rs772945405 Chr.4: 55369981-55369981 on Build GRCh38
rs748009134 Chr.4: 55369969-55369969 on Build GRCh38
rs758311887 Chr.4: 55370128-55370128 on Build GRCh38
rs746726949 Chr.4: 55369985-55369985 on Build GRCh38
rs757565408 Chr.4: 55370061-55370061 on Build GRCh38
rs865889003 Chr.4: 55370014-55370014 on Build GRCh38
rs776147710 Chr.4: 55370040-55370040 on Build GRCh38
rs758956325 Chr.4: 55370028-55370028 on Build GRCh38
rs995935271 Chr.4: 55370112-55370112 on Build GRCh38
rs771882471 Chr.4: 55369971-55369971 on Build GRCh38
rs776275493 Chr.4: 55370023-55370023 on Build GRCh38
rs780065677 Chr.4: 55370107-55370107 on Build GRCh38
rs748291303 Chr.4: 55370103-55370103 on Build GRCh38
rs2538 Chr.4: 55370161-55370161 on Build GRCh38
rs764601564 Chr.4: 55370058-55370058 on Build GRCh38
rs767620528 Chr.4: 55370066-55370066 on Build GRCh38
rs569181314 Chr.4: 55369955-55369955 on Build GRCh38
rs750597249 Chr.4: 55370077-55370077 on Build GRCh38
rs61733673 Chr.4: 55370059-55370059 on Build GRCh38
rs749145324 Chr.4: 55370133-55370133 on Build GRCh38

View Full Product Details