Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56221415-56221676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN5A
Entrez Gene ID
79149
Gene Name
zinc finger and SCAN domain containing 5A
Gene Aliases
ZNF495, ZSCAN5
UniGene
Hs.177688
Gene Chromosome Location
Chr.19: 56221303-56368383 on Build GRCh38
Transcripts
NM_001322061.1 NM_001322062.1 NM_001322064.1 NM_001322065.1 NM_001322066.1 NM_001322067.1 NM_001322068.1 NM_001322069.1 NM_001322070.1 NM_001322072.1 NM_001322073.1 NM_001322074.1 NM_001322075.1 NM_001322076.1 NM_001322077.1 NM_001322078.1 NM_024303.2 XM_005259254.1 XM_006723377.3 XM_011527308.1 XM_017027298.1 XM_017027299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7174777 c.1448T>C p.L483S Chr.19: 56221618-56221618 on Build GRCh38
1293867 c.1402T>C p.Y468H Chr.19: 56221664-56221664 on Build GRCh38
4081766 c.1398A>C p.K466N Chr.19: 56221668-56221668 on Build GRCh38
3539976 c.1391G>A p.G464E Chr.19: 56221675-56221675 on Build GRCh38
475335 c.1491A>G p.*497W Chr.19: 56221575-56221575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779668524 Chr.19: 56221645-56221645 on Build GRCh38
rs754807953 Chr.19: 56221647-56221647 on Build GRCh38
rs150349355 Chr.19: 56221578-56221578 on Build GRCh38
rs764297887 Chr.19: 56221613-56221613 on Build GRCh38
rs367602756 Chr.19: 56221484-56221484 on Build GRCh38
rs148798821 Chr.19: 56221516-56221516 on Build GRCh38
rs753302376 Chr.19: 56221599-56221599 on Build GRCh38
rs138110009 Chr.19: 56221587-56221587 on Build GRCh38
rs536449963 Chr.19: 56221530-56221530 on Build GRCh38
rs142383251 Chr.19: 56221520-56221520 on Build GRCh38
rs867605732 Chr.19: 56221464-56221464 on Build GRCh38
rs772728170 Chr.19: 56221581-56221581 on Build GRCh38
rs765803270 Chr.19: 56221594-56221594 on Build GRCh38
rs758587498 Chr.19: 56221558-56221558 on Build GRCh38
rs752854954 Chr.19: 56221556-56221556 on Build GRCh38
rs939621457 Chr.19: 56221542-56221542 on Build GRCh38
rs199652968 Chr.19: 56221585-56221585 on Build GRCh38
rs376608225 Chr.19: 56221445-56221445 on Build GRCh38
rs181267764 Chr.19: 56221468-56221468 on Build GRCh38
rs1019389882 Chr.19: 56221483-56221483 on Build GRCh38
rs777505765 Chr.19: 56221528-56221529 on Build GRCh38
rs757517666 Chr.19: 56221634-56221634 on Build GRCh38
rs980905918 Chr.19: 56221528-56221530 on Build GRCh38
rs930804885 Chr.19: 56221522-56221522 on Build GRCh38
rs62122488 Chr.19: 56221446-56221446 on Build GRCh38
rs753667989 Chr.19: 56221646-56221646 on Build GRCh38
rs535253175 Chr.19: 56221482-56221482 on Build GRCh38
rs777832426 Chr.19: 56221559-56221559 on Build GRCh38
rs747626844 Chr.19: 56221576-56221576 on Build GRCh38
rs202216893 Chr.19: 56221564-56221564 on Build GRCh38
rs368375280 Chr.19: 56221529-56221529 on Build GRCh38
rs377759902 Chr.19: 56221555-56221555 on Build GRCh38
rs781680429 Chr.19: 56221536-56221538 on Build GRCh38
rs777075627 Chr.19: 56221586-56221586 on Build GRCh38
rs751669053 Chr.19: 56221631-56221631 on Build GRCh38
rs1056705456 Chr.19: 56221543-56221543 on Build GRCh38
rs761391841 Chr.19: 56221565-56221565 on Build GRCh38
rs200882365 Chr.19: 56221650-56221650 on Build GRCh38
rs773810597 Chr.19: 56221570-56221570 on Build GRCh38
rs747310164 Chr.19: 56221562-56221562 on Build GRCh38
rs371193579 Chr.19: 56221563-56221563 on Build GRCh38
rs763109321 Chr.19: 56221612-56221612 on Build GRCh38
rs987509992 Chr.19: 56221470-56221470 on Build GRCh38
rs553702469 Chr.19: 56221444-56221444 on Build GRCh38
rs754229834 Chr.19: 56221531-56221531 on Build GRCh38
rs755554696 Chr.19: 56221554-56221554 on Build GRCh38

View Full Product Details