Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 52085077-52085302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP64
Entrez Gene ID
55734
Gene Name
ZFP64 zinc finger protein
Gene Aliases
ZNF338
UniGene
Hs.473082
Gene Chromosome Location
Chr.20: 52084011-52191985 on Build GRCh38
Transcripts
NM_001319146.1 NM_199427.2 XM_006723822.1 XM_017027945.1 XM_017027947.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6826777 c.1345G>A p.V449I Chr.20: 52085150-52085150 on Build GRCh38
444042 c.1374A>G p.K458K Chr.20: 52085121-52085121 on Build GRCh38
5632746 c.1348C>T p.R450C Chr.20: 52085147-52085147 on Build GRCh38
5631047 c.1280C>T p.S427L Chr.20: 52085215-52085215 on Build GRCh38
6826774 c.727C>T p.R243C Chr.20: 52085111-52085111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764201859 Chr.20: 52085183-52085183 on Build GRCh38
rs777283270 Chr.20: 52085269-52085269 on Build GRCh38
rs934658681 Chr.20: 52085233-52085233 on Build GRCh38
rs770538512 Chr.20: 52085145-52085145 on Build GRCh38
rs760902690 Chr.20: 52085174-52085174 on Build GRCh38
rs374976001 Chr.20: 52085130-52085130 on Build GRCh38
rs755974345 Chr.20: 52085264-52085264 on Build GRCh38
rs760052684 Chr.20: 52085151-52085151 on Build GRCh38
rs376472510 Chr.20: 52085163-52085163 on Build GRCh38
rs369176901 Chr.20: 52085139-52085139 on Build GRCh38
rs767372174 Chr.20: 52085256-52085256 on Build GRCh38
rs765418770 Chr.20: 52085211-52085211 on Build GRCh38
rs757911046 Chr.20: 52085152-52085152 on Build GRCh38
rs749024554 Chr.20: 52085270-52085270 on Build GRCh38
rs751238533 Chr.20: 52085187-52085187 on Build GRCh38
rs752407916 Chr.20: 52085257-52085257 on Build GRCh38
rs776953169 Chr.20: 52085188-52085188 on Build GRCh38
rs560159731 Chr.20: 52085153-52085153 on Build GRCh38
rs754996035 Chr.20: 52085247-52085247 on Build GRCh38
rs751474915 Chr.20: 52085234-52085234 on Build GRCh38
rs373215737 Chr.20: 52085147-52085147 on Build GRCh38
rs532141806 Chr.20: 52085199-52085199 on Build GRCh38

View Full Product Details