Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180114483-180114712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASGEF1C
Entrez Gene ID
255426
Gene Name
RasGEF domain family member 1C
Gene Aliases
-
UniGene
Hs.190559
Gene Chromosome Location
Chr.5: 180100795-180209130 on Build GRCh38
Transcripts
NM_175062.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6878785 c.1114A>G p.K372E Chr.5: 180114511-180114511 on Build GRCh38
3614823 c.1084-1G>A p.? Chr.5: 180114542-180114542 on Build GRCh38
7332605 c.1107G>T p.L369L Chr.5: 180114518-180114518 on Build GRCh38
3614822 c.1084-1G>A p.? Chr.5: 180114542-180114542 on Build GRCh38
5604663 c.1128C>T p.F376F Chr.5: 180114497-180114497 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372677930 Chr.5: 180114575-180114575 on Build GRCh38
rs1032988841 Chr.5: 180114618-180114618 on Build GRCh38
rs775024415 Chr.5: 180114584-180114584 on Build GRCh38
rs1024391419 Chr.5: 180114665-180114665 on Build GRCh38
rs753565208 Chr.5: 180114523-180114524 on Build GRCh38
rs780173570 Chr.5: 180114506-180114506 on Build GRCh38
rs905315449 Chr.5: 180114651-180114651 on Build GRCh38
rs1036297834 Chr.5: 180114602-180114602 on Build GRCh38
rs866637646 Chr.5: 180114680-180114680 on Build GRCh38
rs759083394 Chr.5: 180114574-180114574 on Build GRCh38
rs770990697 Chr.5: 180114569-180114569 on Build GRCh38
rs781242750 Chr.5: 180114556-180114556 on Build GRCh38
rs896482562 Chr.5: 180114624-180114624 on Build GRCh38
rs747076564 Chr.5: 180114518-180114518 on Build GRCh38
rs755036869 Chr.5: 180114551-180114552 on Build GRCh38
rs547819125 Chr.5: 180114589-180114589 on Build GRCh38
rs765216474 Chr.5: 180114648-180114648 on Build GRCh38
rs752511737 Chr.5: 180114692-180114692 on Build GRCh38
rs371249576 Chr.5: 180114562-180114562 on Build GRCh38
rs949464348 Chr.5: 180114640-180114640 on Build GRCh38
rs1037302650 Chr.5: 180114544-180114544 on Build GRCh38
rs759027881 Chr.5: 180114601-180114601 on Build GRCh38
rs755240281 Chr.5: 180114539-180114539 on Build GRCh38
rs749245240 Chr.5: 180114565-180114565 on Build GRCh38
rs1000209646 Chr.5: 180114610-180114610 on Build GRCh38
rs931691088 Chr.5: 180114567-180114567 on Build GRCh38
rs566338876 Chr.5: 180114662-180114662 on Build GRCh38
rs778020989 Chr.5: 180114556-180114557 on Build GRCh38
rs749871504 Chr.5: 180114564-180114564 on Build GRCh38
rs748363945 Chr.5: 180114558-180114558 on Build GRCh38
rs1019717988 Chr.5: 180114553-180114553 on Build GRCh38
rs750782173 Chr.5: 180114590-180114590 on Build GRCh38
rs539677629 Chr.5: 180114673-180114673 on Build GRCh38
rs774065704 Chr.5: 180114570-180114570 on Build GRCh38
rs1045549191 Chr.5: 180114649-180114649 on Build GRCh38
rs981955873 Chr.5: 180114691-180114691 on Build GRCh38
rs903422397 Chr.5: 180114602-180114602 on Build GRCh38
rs557944051 Chr.5: 180114689-180114689 on Build GRCh38
rs760312923 Chr.5: 180114588-180114588 on Build GRCh38
rs538417154 Chr.5: 180114581-180114582 on Build GRCh38

View Full Product Details