Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41308840-41309069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFRP1
Entrez Gene ID
6422
Gene Name
secreted frizzled related protein 1
Gene Aliases
FRP, FRP-1, FRP1, FrzA, SARP2
UniGene
Hs.213424
Gene Chromosome Location
Chr.8: 41261957-41309471 on Build GRCh38
Transcripts
NM_003012.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4650807 c.281C>T p.A94V Chr.8: 41308879-41308879 on Build GRCh38
6776379 c.103G>A p.D35N Chr.8: 41309057-41309057 on Build GRCh38
1624014 c.139C>A p.Q47K Chr.8: 41309021-41309021 on Build GRCh38
4812981 c.290G>A p.W97* Chr.8: 41308870-41308870 on Build GRCh38
5762301 c.102C>T p.Y34Y Chr.8: 41309058-41309058 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374492383 Chr.8: 41308903-41308903 on Build GRCh38
rs759781230 Chr.8: 41308935-41308935 on Build GRCh38
rs747605410 Chr.8: 41309000-41309000 on Build GRCh38
rs866775064 Chr.8: 41308936-41308936 on Build GRCh38
rs372288204 Chr.8: 41308956-41308956 on Build GRCh38
rs764130978 Chr.8: 41309037-41309037 on Build GRCh38
rs749781995 Chr.8: 41309005-41309005 on Build GRCh38
rs1006637962 Chr.8: 41309019-41309019 on Build GRCh38
rs754619436 Chr.8: 41308992-41308992 on Build GRCh38
rs773186629 Chr.8: 41309038-41309038 on Build GRCh38
rs866487203 Chr.8: 41309029-41309029 on Build GRCh38
rs761741033 Chr.8: 41308959-41308959 on Build GRCh38
rs774476008 Chr.8: 41308933-41308933 on Build GRCh38
rs751194586 Chr.8: 41308985-41308985 on Build GRCh38
rs76486417 Chr.8: 41308916-41308916 on Build GRCh38
rs551082706 Chr.8: 41309028-41309028 on Build GRCh38
rs964980693 Chr.8: 41309039-41309039 on Build GRCh38
rs765098694 Chr.8: 41308962-41308962 on Build GRCh38
rs766178906 Chr.8: 41308984-41308984 on Build GRCh38
rs780574104 Chr.8: 41308998-41308998 on Build GRCh38
rs148373821 Chr.8: 41309025-41309025 on Build GRCh38
rs758232055 Chr.8: 41308980-41308980 on Build GRCh38
rs746645058 Chr.8: 41308863-41308863 on Build GRCh38
rs755670568 Chr.8: 41309001-41309001 on Build GRCh38
rs766118970 Chr.8: 41309046-41309046 on Build GRCh38
rs1057451989 Chr.8: 41308891-41308891 on Build GRCh38
rs184528079 Chr.8: 41309004-41309004 on Build GRCh38
rs937435721 Chr.8: 41308880-41308880 on Build GRCh38
rs762832033 Chr.8: 41309045-41309045 on Build GRCh38
rs772267698 Chr.8: 41309026-41309026 on Build GRCh38
rs1016241797 Chr.8: 41308947-41308947 on Build GRCh38
rs773726937 Chr.8: 41308907-41308907 on Build GRCh38
rs767514199 Chr.8: 41308942-41308942 on Build GRCh38
rs768103490 Chr.8: 41308876-41308876 on Build GRCh38
rs768472530 Chr.8: 41309049-41309049 on Build GRCh38
rs780705476 Chr.8: 41308878-41308878 on Build GRCh38
rs748698728 Chr.8: 41308892-41308892 on Build GRCh38
rs750358294 Chr.8: 41308967-41308967 on Build GRCh38
rs141546086 Chr.8: 41309047-41309047 on Build GRCh38
rs771249090 Chr.8: 41309011-41309011 on Build GRCh38

View Full Product Details