Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 107311491-107311748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD23B
Entrez Gene ID
5887
Gene Name
RAD23 homolog B, nucleotide excision repair protein
Gene Aliases
HHR23B, HR23B, P58
UniGene
Hs.521640
Gene Chromosome Location
Chr.9: 107283236-107332194 on Build GRCh38
Transcripts
NM_001244713.1 NM_001244724.1 NM_002874.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5947088 c.162+1_162+4delGTAA p.? Chr.9: 107311738-107311741 on Build GRCh38
3375029 c.162+1G>T p.? Chr.9: 107311738-107311738 on Build GRCh38
140627 c.508G>A p.D170N Chr.9: 107311692-107311692 on Build GRCh38
6182119 c.150A>G p.A50A Chr.9: 107311725-107311725 on Build GRCh38
5947087 c.553+1_553+4delGTAA p.? Chr.9: 107311738-107311741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759438239 Chr.9: 107311696-107311696 on Build GRCh38
rs369759634 Chr.9: 107311638-107311638 on Build GRCh38
rs887044942 Chr.9: 107311535-107311535 on Build GRCh38
rs766344267 Chr.9: 107311637-107311637 on Build GRCh38
rs547086698 Chr.9: 107311651-107311651 on Build GRCh38
rs764157337 Chr.9: 107311701-107311701 on Build GRCh38
rs969310129 Chr.9: 107311546-107311546 on Build GRCh38
rs772390908 Chr.9: 107311660-107311660 on Build GRCh38
rs561535881 Chr.9: 107311641-107311641 on Build GRCh38
rs768255578 Chr.9: 107311667-107311667 on Build GRCh38
rs11573678 Chr.9: 107311577-107311577 on Build GRCh38
rs756457087 Chr.9: 107311640-107311640 on Build GRCh38
rs373904260 Chr.9: 107311678-107311678 on Build GRCh38
rs76169041 Chr.9: 107311694-107311694 on Build GRCh38
rs571290306 Chr.9: 107311665-107311665 on Build GRCh38
rs370202368 Chr.9: 107311673-107311673 on Build GRCh38
rs371974204 Chr.9: 107311655-107311655 on Build GRCh38
rs940337929 Chr.9: 107311609-107311609 on Build GRCh38
rs757276908 Chr.9: 107311712-107311712 on Build GRCh38
rs757368676 Chr.9: 107311654-107311654 on Build GRCh38
rs903230511 Chr.9: 107311688-107311688 on Build GRCh38
rs1036139409 Chr.9: 107311635-107311635 on Build GRCh38
rs1013467651 Chr.9: 107311560-107311560 on Build GRCh38
rs766258521 Chr.9: 107311693-107311693 on Build GRCh38
rs763025606 Chr.9: 107311690-107311690 on Build GRCh38
rs747395460 Chr.9: 107311666-107311666 on Build GRCh38
rs754145429 Chr.9: 107311707-107311707 on Build GRCh38
rs758255202 Chr.9: 107311633-107311633 on Build GRCh38
rs908914854 Chr.9: 107311604-107311604 on Build GRCh38
rs189555517 Chr.9: 107311687-107311687 on Build GRCh38
rs528550040 Chr.9: 107311644-107311644 on Build GRCh38

View Full Product Details