Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 99987411-99987654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS22
Entrez Gene ID
26166
Gene Name
regulator of G-protein signaling 22
Gene Aliases
CT145, PRTD-NY2
UniGene
Hs.120021
Gene Chromosome Location
Chr.8: 99960936-100106116 on Build GRCh38
Transcripts
NM_001286692.1 NM_001286693.1 NM_015668.4 XM_005250856.2 XM_005250857.2 XM_005250858.2 XM_006716540.2 XM_011516957.1 XM_011516958.1 XM_017013309.1 XM_017013310.1 XM_017013311.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
400706 c.3168A>T p.V1056V Chr.8: 99987470-99987470 on Build GRCh38
5582341 c.3057C>T p.I1019I Chr.8: 99987581-99987581 on Build GRCh38
3884096 c.3039C>A p.I1013I Chr.8: 99987599-99987599 on Build GRCh38
72420 c.3110G>T p.R1037L Chr.8: 99987528-99987528 on Build GRCh38
6411709 c.3118G>T p.A1040S Chr.8: 99987520-99987520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370688344 Chr.8: 99987494-99987494 on Build GRCh38
rs776254718 Chr.8: 99987499-99987499 on Build GRCh38
rs370094087 Chr.8: 99987446-99987446 on Build GRCh38
rs373414660 Chr.8: 99987449-99987449 on Build GRCh38
rs376301062 Chr.8: 99987487-99987487 on Build GRCh38
rs980175532 Chr.8: 99987506-99987506 on Build GRCh38
rs772757723 Chr.8: 99987622-99987622 on Build GRCh38
rs367712644 Chr.8: 99987570-99987570 on Build GRCh38
rs745674176 Chr.8: 99987462-99987462 on Build GRCh38
rs76193261 Chr.8: 99987440-99987440 on Build GRCh38
rs753963357 Chr.8: 99987538-99987538 on Build GRCh38
rs374356639 Chr.8: 99987536-99987536 on Build GRCh38
rs768439818 Chr.8: 99987588-99987588 on Build GRCh38
rs760075875 Chr.8: 99987511-99987511 on Build GRCh38
rs746755199 Chr.8: 99987586-99987586 on Build GRCh38
rs1008272832 Chr.8: 99987452-99987452 on Build GRCh38
rs757215620 Chr.8: 99987565-99987565 on Build GRCh38
rs771700896 Chr.8: 99987473-99987473 on Build GRCh38
rs745847509 Chr.8: 99987571-99987571 on Build GRCh38
rs932790773 Chr.8: 99987548-99987548 on Build GRCh38
rs780031194 Chr.8: 99987576-99987576 on Build GRCh38
rs982846019 Chr.8: 99987458-99987458 on Build GRCh38
rs756007965 Chr.8: 99987492-99987492 on Build GRCh38
rs373056041 Chr.8: 99987503-99987503 on Build GRCh38
rs952438726 Chr.8: 99987497-99987497 on Build GRCh38
rs182707509 Chr.8: 99987528-99987528 on Build GRCh38
rs769248020 Chr.8: 99987619-99987619 on Build GRCh38
rs775442553 Chr.8: 99987485-99987485 on Build GRCh38
rs764023368 Chr.8: 99987518-99987518 on Build GRCh38
rs199838743 Chr.8: 99987512-99987512 on Build GRCh38
rs774215914 Chr.8: 99987450-99987450 on Build GRCh38
rs761173752 Chr.8: 99987502-99987502 on Build GRCh38
rs200406131 Chr.8: 99987508-99987508 on Build GRCh38
rs113137742 Chr.8: 99987434-99987434 on Build GRCh38
rs761524888 Chr.8: 99987602-99987602 on Build GRCh38
rs760001306 Chr.8: 99987624-99987624 on Build GRCh38
rs370385204 Chr.8: 99987529-99987529 on Build GRCh38
rs772020361 Chr.8: 99987575-99987575 on Build GRCh38
rs776359550 Chr.8: 99987591-99987591 on Build GRCh38

View Full Product Details