Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113157463-113157736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP73
Entrez Gene ID
387885
Gene Name
cilia and flagella associated protein 73
Gene Aliases
CCDC42B, MIA2
UniGene
Hs.486856
Gene Chromosome Location
Chr.12: 113149162-113159276 on Build GRCh38
Transcripts
NM_001144872.1 NM_001111322.1 NM_024072.3

Gene Symbol
DDX54
Entrez Gene ID
79039
Gene Name
DEAD-box helicase 54
Gene Aliases
DP97
UniGene
Hs.506861
Gene Chromosome Location
Chr.12: 113157173-113185479 on Build GRCh38
Transcripts
NM_001144872.1 NM_001111322.1 NM_024072.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7280236 c.850-9delC p.? Chr.12: 113157593-113157593 on Build GRCh38
3811096 c.851T>G p.V284G Chr.12: 113157603-113157603 on Build GRCh38
4953301 c.907C>A p.P303T Chr.12: 113157659-113157659 on Build GRCh38
3687971 c.855G>T p.K285N Chr.12: 113157607-113157607 on Build GRCh38
5781610 c.889G>A p.G297S Chr.12: 113157641-113157641 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768984396 Chr.12: 113157682-113157682 on Build GRCh38
rs913490083 Chr.12: 113157567-113157567 on Build GRCh38
rs746212525 Chr.12: 113157509-113157509 on Build GRCh38
rs373975214 Chr.12: 113157695-113157695 on Build GRCh38
rs896129054 Chr.12: 113157633-113157633 on Build GRCh38
rs917885557 Chr.12: 113157533-113157533 on Build GRCh38
rs769242183 Chr.12: 113157589-113157589 on Build GRCh38
rs936528877 Chr.12: 113157603-113157603 on Build GRCh38
rs1015528370 Chr.12: 113157501-113157501 on Build GRCh38
rs201608216 Chr.12: 113157503-113157504 on Build GRCh38
rs556859691 Chr.12: 113157532-113157532 on Build GRCh38
rs769885042 Chr.12: 113157592-113157593 on Build GRCh38
rs561707496 Chr.12: 113157699-113157699 on Build GRCh38
rs570675069 Chr.12: 113157599-113157599 on Build GRCh38
rs766861972 Chr.12: 113157668-113157671 on Build GRCh38
rs539276285 Chr.12: 113157594-113157594 on Build GRCh38
rs573179592 Chr.12: 113157658-113157658 on Build GRCh38
rs575422183 Chr.12: 113157572-113157572 on Build GRCh38
rs779293026 Chr.12: 113157499-113157499 on Build GRCh38
rs772772117 Chr.12: 113157601-113157601 on Build GRCh38
rs141204840 Chr.12: 113157516-113157516 on Build GRCh38
rs897103294 Chr.12: 113157679-113157679 on Build GRCh38
rs61941412 Chr.12: 113157521-113157521 on Build GRCh38
rs962901492 Chr.12: 113157520-113157520 on Build GRCh38
rs191674171 Chr.12: 113157531-113157531 on Build GRCh38
rs1038837882 Chr.12: 113157630-113157630 on Build GRCh38
rs776776545 Chr.12: 113157690-113157690 on Build GRCh38
rs748939865 Chr.12: 113157593-113157593 on Build GRCh38
rs1013950068 Chr.12: 113157496-113157496 on Build GRCh38
rs1034363109 Chr.12: 113157522-113157522 on Build GRCh38
rs921853546 Chr.12: 113157640-113157640 on Build GRCh38
rs747703497 Chr.12: 113157539-113157539 on Build GRCh38
rs768274814 Chr.12: 113157584-113157584 on Build GRCh38
rs763023498 Chr.12: 113157641-113157641 on Build GRCh38
rs989446989 Chr.12: 113157565-113157566 on Build GRCh38
rs137861202 Chr.12: 113157714-113157715 on Build GRCh38
rs1004676877 Chr.12: 113157510-113157510 on Build GRCh38
rs1037322882 Chr.12: 113157678-113157678 on Build GRCh38

View Full Product Details