Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 46367234-46367483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKZ
Entrez Gene ID
8525
Gene Name
diacylglycerol kinase zeta
Gene Aliases
DAGK5, DAGK6, DGK-ZETA, hDGKzeta
UniGene
Hs.502461
Gene Chromosome Location
Chr.11: 46332905-46380554 on Build GRCh38
Transcripts
NM_001105540.1 NM_001199266.1 NM_001199267.1 NM_001199268.1 NM_003646.3 NM_201532.2 NM_201533.3 XM_005253181.3 XM_005253182.3 XM_006718354.1 XM_006718355.2 XM_011520422.1 XM_011520423.1 XM_017018455.1 XM_017018456.1 XM_017018457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5473132 c.235T>C p.S79P Chr.11: 46367364-46367364 on Build GRCh38
5360861 c.253C>A p.P85T Chr.11: 46367331-46367331 on Build GRCh38
5970169 c.824C>T p.T275I Chr.11: 46367386-46367386 on Build GRCh38
3447536 c.110C>T p.T37I Chr.11: 46367341-46367341 on Build GRCh38
3447530 c.252C>A p.P84P Chr.11: 46367330-46367330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562334771 Chr.11: 46367460-46367460 on Build GRCh38
rs749509532 Chr.11: 46367353-46367353 on Build GRCh38
rs372116043 Chr.11: 46367323-46367323 on Build GRCh38
rs149537688 Chr.11: 46367342-46367342 on Build GRCh38
rs763360462 Chr.11: 46367263-46367263 on Build GRCh38
rs775057820 Chr.11: 46367318-46367318 on Build GRCh38
rs370912270 Chr.11: 46367421-46367421 on Build GRCh38
rs143851594 Chr.11: 46367363-46367363 on Build GRCh38
rs778867449 Chr.11: 46367294-46367294 on Build GRCh38
rs764807844 Chr.11: 46367379-46367379 on Build GRCh38
rs1033888459 Chr.11: 46367438-46367438 on Build GRCh38
rs778188259 Chr.11: 46367351-46367351 on Build GRCh38
rs185575530 Chr.11: 46367439-46367439 on Build GRCh38
rs772181855 Chr.11: 46367340-46367340 on Build GRCh38
rs752166288 Chr.11: 46367327-46367327 on Build GRCh38
rs749314563 Chr.11: 46367434-46367434 on Build GRCh38
rs758278027 Chr.11: 46367336-46367336 on Build GRCh38
rs1057061592 Chr.11: 46367406-46367406 on Build GRCh38
rs761454646 Chr.11: 46367378-46367378 on Build GRCh38
rs763578474 Chr.11: 46367371-46367371 on Build GRCh38
rs1041225858 Chr.11: 46367345-46367345 on Build GRCh38
rs375162900 Chr.11: 46367370-46367370 on Build GRCh38
rs554583193 Chr.11: 46367313-46367313 on Build GRCh38
rs147007748 Chr.11: 46367331-46367331 on Build GRCh38
rs755730987 Chr.11: 46367271-46367271 on Build GRCh38
rs777998831 Chr.11: 46367432-46367432 on Build GRCh38
rs781062257 Chr.11: 46367280-46367282 on Build GRCh38
rs372222252 Chr.11: 46367278-46367278 on Build GRCh38
rs759891562 Chr.11: 46367361-46367361 on Build GRCh38
rs755605885 Chr.11: 46367329-46367329 on Build GRCh38
rs753553170 Chr.11: 46367374-46367374 on Build GRCh38
rs1024824543 Chr.11: 46367462-46367462 on Build GRCh38
rs759956247 Chr.11: 46367320-46367320 on Build GRCh38
rs894294749 Chr.11: 46367262-46367262 on Build GRCh38
rs748200605 Chr.11: 46367306-46367306 on Build GRCh38
rs758172144 Chr.11: 46367386-46367386 on Build GRCh38
rs774861524 Chr.11: 46367357-46367357 on Build GRCh38
rs777352534 Chr.11: 46367275-46367275 on Build GRCh38
rs367575196 Chr.11: 46367280-46367280 on Build GRCh38
rs754568079 Chr.11: 46367265-46367265 on Build GRCh38
rs766923753 Chr.11: 46367267-46367267 on Build GRCh38
rs576632084 Chr.11: 46367425-46367425 on Build GRCh38
rs748207784 Chr.11: 46367341-46367341 on Build GRCh38
rs539478353 Chr.11: 46367275-46367277 on Build GRCh38
rs745652061 Chr.11: 46367325-46367326 on Build GRCh38
rs369496955 Chr.11: 46367298-46367298 on Build GRCh38
rs773724906 Chr.11: 46367261-46367261 on Build GRCh38
rs1013269703 Chr.11: 46367453-46367453 on Build GRCh38
rs961264855 Chr.11: 46367464-46367464 on Build GRCh38
rs141368349 Chr.11: 46367309-46367309 on Build GRCh38
rs138240327 Chr.11: 46367335-46367335 on Build GRCh38
rs148198310 Chr.11: 46367366-46367366 on Build GRCh38
rs568978655 Chr.11: 46367308-46367308 on Build GRCh38
rs3740978 Chr.11: 46367332-46367332 on Build GRCh38
rs141189455 Chr.11: 46367383-46367383 on Build GRCh38
rs779893573 Chr.11: 46367420-46367420 on Build GRCh38
rs776575197 Chr.11: 46367305-46367305 on Build GRCh38
rs138804845 Chr.11: 46367300-46367300 on Build GRCh38
rs17854148 Chr.11: 46367303-46367303 on Build GRCh38
rs767022649 Chr.11: 46367264-46367264 on Build GRCh38

View Full Product Details