Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 87072751-87073024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS2ST1
Entrez Gene ID
9653
Gene Name
heparan sulfate 2-O-sulfotransferase 1
Gene Aliases
dJ604K5.2
UniGene
Hs.743280
Gene Chromosome Location
Chr.1: 86914652-87109998 on Build GRCh38
Transcripts
NM_001134492.1 NM_012262.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7199381 c.47A>G p.E16G Chr.1: 87072934-87072934 on Build GRCh38
1748636 c.187C>T p.R63W Chr.1: 87072996-87072996 on Build GRCh38
168989 c.151C>T p.R51* Chr.1: 87072960-87072960 on Build GRCh38
7278033 c.172A>G p.T58A Chr.1: 87072981-87072981 on Build GRCh38
1748637 c.109C>T p.R37W Chr.1: 87072996-87072996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763680605 Chr.1: 87072953-87072953 on Build GRCh38
rs368496293 Chr.1: 87072996-87072996 on Build GRCh38
rs147874594 Chr.1: 87072997-87072997 on Build GRCh38
rs775025973 Chr.1: 87072943-87072943 on Build GRCh38
rs964522001 Chr.1: 87072801-87072801 on Build GRCh38
rs891426450 Chr.1: 87072824-87072825 on Build GRCh38
rs1008745752 Chr.1: 87072782-87072782 on Build GRCh38
rs370721303 Chr.1: 87072917-87072917 on Build GRCh38
rs35522446 Chr.1: 87072898-87072899 on Build GRCh38
rs761491625 Chr.1: 87072969-87072969 on Build GRCh38
rs983091583 Chr.1: 87072952-87072952 on Build GRCh38
rs964583821 Chr.1: 87072883-87072883 on Build GRCh38
rs762375562 Chr.1: 87072945-87072945 on Build GRCh38
rs566716808 Chr.1: 87072867-87072867 on Build GRCh38
rs760225543 Chr.1: 87072780-87072780 on Build GRCh38
rs771094744 Chr.1: 87072784-87072784 on Build GRCh38
rs756054488 Chr.1: 87072902-87072902 on Build GRCh38
rs767275935 Chr.1: 87072976-87072976 on Build GRCh38
rs865888400 Chr.1: 87072960-87072960 on Build GRCh38
rs986791348 Chr.1: 87072966-87072966 on Build GRCh38
rs187981521 Chr.1: 87072859-87072859 on Build GRCh38
rs750206376 Chr.1: 87072981-87072981 on Build GRCh38
rs1017884774 Chr.1: 87072799-87072799 on Build GRCh38
rs771420041 Chr.1: 87072927-87072927 on Build GRCh38
rs376848829 Chr.1: 87072914-87072914 on Build GRCh38
rs746367272 Chr.1: 87072935-87072935 on Build GRCh38
rs370053454 Chr.1: 87072992-87072992 on Build GRCh38
rs773981936 Chr.1: 87072954-87072954 on Build GRCh38
rs376924859 Chr.1: 87072987-87072987 on Build GRCh38
rs777179312 Chr.1: 87072931-87072931 on Build GRCh38
rs202115672 Chr.1: 87072913-87072913 on Build GRCh38
rs147264643 Chr.1: 87072942-87072942 on Build GRCh38
rs1011343622 Chr.1: 87072836-87072836 on Build GRCh38
rs747290455 Chr.1: 87072921-87072921 on Build GRCh38
rs760621318 Chr.1: 87072984-87072984 on Build GRCh38

View Full Product Details