Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80521860-80522101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM151B
Entrez Gene ID
167555
Gene Name
family with sequence similarity 151 member B
Gene Aliases
UNQ9217
UniGene
Hs.338182
Gene Chromosome Location
Chr.5: 80487981-80542563 on Build GRCh38
Transcripts
NM_205548.2 XM_006714564.3 XM_011543234.2 XM_011543235.2 XM_011543236.2 XM_011543237.2 XM_017009166.1 XM_017009167.1 XM_017009168.1 XM_017009169.1 XM_017009170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
369682 c.595G>T p.V199L Chr.5: 80522062-80522062 on Build GRCh38
4682617 c.567_568delAT p.I191fs*2 Chr.5: 80522034-80522035 on Build GRCh38
1438880 c.609C>T p.V203V Chr.5: 80522076-80522076 on Build GRCh38
7193410 c.633T>G p.S211S Chr.5: 80522100-80522100 on Build GRCh38
7076952 c.583C>A p.L195I Chr.5: 80522050-80522050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201044459 Chr.5: 80521957-80521957 on Build GRCh38
rs1008779259 Chr.5: 80522062-80522062 on Build GRCh38
rs578022977 Chr.5: 80522042-80522042 on Build GRCh38
rs555386964 Chr.5: 80521895-80521895 on Build GRCh38
rs775123191 Chr.5: 80522070-80522070 on Build GRCh38
rs776106459 Chr.5: 80522040-80522040 on Build GRCh38
rs781216313 Chr.5: 80521985-80521985 on Build GRCh38
rs201474416 Chr.5: 80521974-80521974 on Build GRCh38
rs745986240 Chr.5: 80521990-80521990 on Build GRCh38
rs755735123 Chr.5: 80522053-80522056 on Build GRCh38
rs1053112166 Chr.5: 80522022-80522022 on Build GRCh38
rs756110546 Chr.5: 80521995-80521995 on Build GRCh38
rs1051233543 Chr.5: 80522012-80522012 on Build GRCh38
rs992010876 Chr.5: 80521919-80521919 on Build GRCh38
rs764698345 Chr.5: 80522059-80522059 on Build GRCh38
rs983208604 Chr.5: 80521968-80521968 on Build GRCh38
rs748133725 Chr.5: 80522000-80522000 on Build GRCh38
rs200357393 Chr.5: 80522008-80522008 on Build GRCh38
rs908515372 Chr.5: 80521972-80521972 on Build GRCh38
rs2431224 Chr.5: 80522079-80522079 on Build GRCh38
rs534823766 Chr.5: 80521958-80521958 on Build GRCh38
rs770638463 Chr.5: 80522032-80522032 on Build GRCh38
rs746747618 Chr.5: 80522009-80522009 on Build GRCh38
rs764607719 Chr.5: 80521965-80521965 on Build GRCh38
rs34930902 Chr.5: 80522069-80522070 on Build GRCh38
rs891389030 Chr.5: 80522035-80522035 on Build GRCh38
rs945619450 Chr.5: 80522066-80522066 on Build GRCh38
rs916274657 Chr.5: 80521929-80521929 on Build GRCh38
rs780155460 Chr.5: 80521996-80521996 on Build GRCh38
rs757555759 Chr.5: 80521982-80521982 on Build GRCh38
rs763618213 Chr.5: 80522074-80522074 on Build GRCh38
rs779639831 Chr.5: 80522037-80522037 on Build GRCh38

View Full Product Details