Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6471241-6471484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND1C
Entrez Gene ID
79958
Gene Name
DENN domain containing 1C
Gene Aliases
FAM31C
UniGene
Hs.236449
Gene Chromosome Location
Chr.19: 6467204-6481808 on Build GRCh38
Transcripts
NM_001290331.1 NM_024898.3 XM_006722905.2 XM_006722906.3 XM_011528318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6839724 c.1347G>C p.Q449H Chr.19: 6471286-6471286 on Build GRCh38
1397382 c.1215C>T p.F405F Chr.19: 6471440-6471440 on Build GRCh38
4676975 c.1205delC p.S402fs*1 Chr.19: 6471450-6471450 on Build GRCh38
4830046 c.1340C>T p.S447F Chr.19: 6471293-6471293 on Build GRCh38
4600286 c.1203C>G p.F401L Chr.19: 6471452-6471452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561041826 Chr.19: 6471344-6471344 on Build GRCh38
rs761187263 Chr.19: 6471440-6471440 on Build GRCh38
rs748073483 Chr.19: 6471315-6471315 on Build GRCh38
rs12610646 Chr.19: 6471349-6471349 on Build GRCh38
rs530633389 Chr.19: 6471285-6471285 on Build GRCh38
rs1021313932 Chr.19: 6471389-6471389 on Build GRCh38
rs934262031 Chr.19: 6471296-6471296 on Build GRCh38
rs201113925 Chr.19: 6471418-6471418 on Build GRCh38
rs774049865 Chr.19: 6471335-6471335 on Build GRCh38
rs765609470 Chr.19: 6471461-6471461 on Build GRCh38
rs200123826 Chr.19: 6471280-6471280 on Build GRCh38
rs764648013 Chr.19: 6471454-6471454 on Build GRCh38
rs771515202 Chr.19: 6471422-6471422 on Build GRCh38
rs776469771 Chr.19: 6471311-6471311 on Build GRCh38
rs376706867 Chr.19: 6471450-6471450 on Build GRCh38
rs544452638 Chr.19: 6471293-6471293 on Build GRCh38
rs904954828 Chr.19: 6471375-6471375 on Build GRCh38
rs746068147 Chr.19: 6471378-6471378 on Build GRCh38
rs765889849 Chr.19: 6471323-6471323 on Build GRCh38
rs371502122 Chr.19: 6471276-6471276 on Build GRCh38
rs774558415 Chr.19: 6471431-6471433 on Build GRCh38
rs756576790 Chr.19: 6471357-6471357 on Build GRCh38
rs777723296 Chr.19: 6471410-6471410 on Build GRCh38
rs764482645 Chr.19: 6471315-6471315 on Build GRCh38
rs530060301 Chr.19: 6471398-6471398 on Build GRCh38
rs772184055 Chr.19: 6471304-6471304 on Build GRCh38
rs769732407 Chr.19: 6471318-6471318 on Build GRCh38
rs758454270 Chr.19: 6471383-6471383 on Build GRCh38
rs755657749 Chr.19: 6471322-6471322 on Build GRCh38
rs774917252 Chr.19: 6471431-6471431 on Build GRCh38
rs780230613 Chr.19: 6471386-6471386 on Build GRCh38
rs767033476 Chr.19: 6471347-6471347 on Build GRCh38
rs745988211 Chr.19: 6471297-6471297 on Build GRCh38
rs749104184 Chr.19: 6471415-6471415 on Build GRCh38
rs777282605 Chr.19: 6471336-6471336 on Build GRCh38
rs754147257 Chr.19: 6471367-6471367 on Build GRCh38
rs139694124 Chr.19: 6471270-6471270 on Build GRCh38
rs748126402 Chr.19: 6471387-6471387 on Build GRCh38
rs779066278 Chr.19: 6471372-6471372 on Build GRCh38
rs753228141 Chr.19: 6471355-6471355 on Build GRCh38
rs759180582 Chr.19: 6471419-6471419 on Build GRCh38
rs762698755 Chr.19: 6471320-6471320 on Build GRCh38
rs757632599 Chr.19: 6471368-6471368 on Build GRCh38
rs1030071474 Chr.19: 6471371-6471371 on Build GRCh38
rs747836090 Chr.19: 6471277-6471277 on Build GRCh38
rs537937769 Chr.19: 6471456-6471456 on Build GRCh38

View Full Product Details