Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78630941-78631190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB4
Entrez Gene ID
1143
Gene Name
cholinergic receptor nicotinic beta 4 subunit
Gene Aliases
-
UniGene
Hs.624178
Gene Chromosome Location
Chr.15: 78623282-78655586 on Build GRCh38
Transcripts
NM_000750.3 NM_001256567.1 XM_011521186.2 XM_011521187.2 XM_011521190.2 XM_011521191.2 XM_011521192.2 XM_017021885.1 XM_017021886.1 XM_017021887.1 XM_017021888.1 XM_017021889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2011234 c.295G>A p.G99S Chr.15: 78631140-78631140 on Build GRCh38
5567005 c.266G>A p.R89H Chr.15: 78631169-78631169 on Build GRCh38
5966570 c.296G>T p.G99V Chr.15: 78631139-78631139 on Build GRCh38
6628790 c.324G>T p.K108N Chr.15: 78631111-78631111 on Build GRCh38
965420 c.348G>A p.V116V Chr.15: 78631087-78631087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141056480 Chr.15: 78631090-78631090 on Build GRCh38
rs926514921 Chr.15: 78631064-78631064 on Build GRCh38
rs368556929 Chr.15: 78631025-78631025 on Build GRCh38
rs572877281 Chr.15: 78631007-78631007 on Build GRCh38
rs772727413 Chr.15: 78631148-78631148 on Build GRCh38
rs753591873 Chr.15: 78631062-78631062 on Build GRCh38
rs746284415 Chr.15: 78631110-78631110 on Build GRCh38
rs1041239594 Chr.15: 78631112-78631112 on Build GRCh38
rs770178895 Chr.15: 78631120-78631120 on Build GRCh38
rs879114631 Chr.15: 78631153-78631153 on Build GRCh38
rs750397056 Chr.15: 78631024-78631024 on Build GRCh38
rs766251392 Chr.15: 78631036-78631036 on Build GRCh38
rs952930498 Chr.15: 78631070-78631070 on Build GRCh38
rs747045440 Chr.15: 78631104-78631104 on Build GRCh38
rs1035690644 Chr.15: 78630992-78630992 on Build GRCh38
rs201464909 Chr.15: 78631073-78631073 on Build GRCh38
rs915973505 Chr.15: 78631017-78631017 on Build GRCh38
rs777970629 Chr.15: 78631093-78631093 on Build GRCh38
rs370090528 Chr.15: 78631034-78631034 on Build GRCh38
rs199599011 Chr.15: 78631089-78631089 on Build GRCh38
rs769692180 Chr.15: 78631081-78631081 on Build GRCh38
rs773923621 Chr.15: 78631139-78631139 on Build GRCh38
rs771669915 Chr.15: 78631144-78631144 on Build GRCh38
rs759310176 Chr.15: 78631169-78631169 on Build GRCh38
rs949511652 Chr.15: 78631031-78631031 on Build GRCh38
rs960029752 Chr.15: 78630993-78630993 on Build GRCh38
rs149832833 Chr.15: 78631109-78631109 on Build GRCh38
rs373299350 Chr.15: 78631149-78631149 on Build GRCh38
rs12914008 Chr.15: 78631163-78631163 on Build GRCh38
rs561683947 Chr.15: 78631141-78631141 on Build GRCh38
rs746129925 Chr.15: 78630988-78630988 on Build GRCh38

View Full Product Details