Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 46138377-46138610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTCD
Entrez Gene ID
10841
Gene Name
formimidoyltransferase cyclodeaminase
Gene Aliases
LCHC1
UniGene
Hs.415846
Gene Chromosome Location
Chr.21: 46135981-46156481 on Build GRCh38
Transcripts
NM_001320412.1 NM_006657.2 NM_206965.1 XM_006723961.3 XM_006723962.3 XM_011529434.2 XM_011529435.2 XM_011529436.2 XM_011529437.2 XM_011529439.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6233611 c.1405G>A p.A469T Chr.21: 46138546-46138546 on Build GRCh38
4685791 c.1391C>T p.A464V Chr.21: 46138560-46138560 on Build GRCh38
4955641 c.1409G>A p.R470Q Chr.21: 46138542-46138542 on Build GRCh38
4525990 c.1368G>A p.E456E Chr.21: 46138583-46138583 on Build GRCh38
6233612 c.1405G>A p.A469T Chr.21: 46138546-46138546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762273298 Chr.21: 46138495-46138495 on Build GRCh38
rs369638970 Chr.21: 46138592-46138592 on Build GRCh38
rs763159647 Chr.21: 46138483-46138483 on Build GRCh38
rs779682626 Chr.21: 46138574-46138574 on Build GRCh38
rs764143379 Chr.21: 46138489-46138489 on Build GRCh38
rs762727268 Chr.21: 46138561-46138561 on Build GRCh38
rs771467051 Chr.21: 46138431-46138431 on Build GRCh38
rs61735840 Chr.21: 46138587-46138587 on Build GRCh38
rs780933856 Chr.21: 46138526-46138526 on Build GRCh38
rs566769020 Chr.21: 46138410-46138410 on Build GRCh38
rs568872542 Chr.21: 46138527-46138527 on Build GRCh38
rs544253281 Chr.21: 46138580-46138580 on Build GRCh38
rs771378909 Chr.21: 46138498-46138499 on Build GRCh38
rs773933014 Chr.21: 46138586-46138586 on Build GRCh38
rs767969299 Chr.21: 46138496-46138496 on Build GRCh38
rs749447695 Chr.21: 46138541-46138541 on Build GRCh38
rs576877652 Chr.21: 46138562-46138562 on Build GRCh38
rs775837925 Chr.21: 46138532-46138532 on Build GRCh38
rs756372120 Chr.21: 46138500-46138500 on Build GRCh38
rs755986536 Chr.21: 46138572-46138572 on Build GRCh38
rs773794646 Chr.21: 46138491-46138491 on Build GRCh38
rs750312666 Chr.21: 46138571-46138571 on Build GRCh38
rs765010204 Chr.21: 46138504-46138504 on Build GRCh38
rs749039145 Chr.21: 46138575-46138575 on Build GRCh38
rs372045405 Chr.21: 46138499-46138499 on Build GRCh38
rs777099958 Chr.21: 46138584-46138585 on Build GRCh38
rs962718994 Chr.21: 46138458-46138458 on Build GRCh38
rs61735839 Chr.21: 46138542-46138542 on Build GRCh38
rs978258015 Chr.21: 46138591-46138591 on Build GRCh38
rs571676774 Chr.21: 46138514-46138514 on Build GRCh38
rs556952105 Chr.21: 46138522-46138522 on Build GRCh38
rs188377424 Chr.21: 46138560-46138560 on Build GRCh38
rs774886583 Chr.21: 46138492-46138492 on Build GRCh38
rs750817590 Chr.21: 46138499-46138499 on Build GRCh38
rs776601046 Chr.21: 46138473-46138473 on Build GRCh38
rs779047080 Chr.21: 46138581-46138581 on Build GRCh38
rs908418845 Chr.21: 46138452-46138452 on Build GRCh38
rs536554400 Chr.21: 46138531-46138531 on Build GRCh38
rs757284370 Chr.21: 46138523-46138523 on Build GRCh38
rs756761574 Chr.21: 46138564-46138564 on Build GRCh38
rs775507481 Chr.21: 46138475-46138475 on Build GRCh38
rs977971213 Chr.21: 46138476-46138476 on Build GRCh38
rs200283734 Chr.21: 46138593-46138593 on Build GRCh38
rs539095795 Chr.21: 46138521-46138521 on Build GRCh38
rs781255845 Chr.21: 46138568-46138568 on Build GRCh38
rs199807194 Chr.21: 46138543-46138543 on Build GRCh38
rs771613672 Chr.21: 46138461-46138462 on Build GRCh38
rs527534248 Chr.21: 46138430-46138430 on Build GRCh38
rs368782658 Chr.21: 46138563-46138563 on Build GRCh38
rs552332184 Chr.21: 46138437-46138437 on Build GRCh38
rs773644251 Chr.21: 46138474-46138474 on Build GRCh38
rs976876790 Chr.21: 46138472-46138472 on Build GRCh38
rs1047179 Chr.21: 46138559-46138559 on Build GRCh38
rs761059490 Chr.21: 46138590-46138590 on Build GRCh38
rs376572763 Chr.21: 46138588-46138588 on Build GRCh38
rs773590831 Chr.21: 46138550-46138550 on Build GRCh38
rs374170977 Chr.21: 46138546-46138546 on Build GRCh38
rs921248056 Chr.21: 46138490-46138491 on Build GRCh38
rs752283011 Chr.21: 46138505-46138505 on Build GRCh38
rs904084233 Chr.21: 46138503-46138503 on Build GRCh38

View Full Product Details