Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120750472-120750733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2E1
Entrez Gene ID
2960
Gene Name
general transcription factor IIE subunit 1
Gene Aliases
FE, TF2E1, TFIIE-A
UniGene
Hs.445272
Gene Chromosome Location
Chr.3: 120742711-120783134 on Build GRCh38
Transcripts
NM_005513.2 XM_011512744.2 XM_011512745.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1036871 c.166C>T p.R56W Chr.3: 120750718-120750718 on Build GRCh38
6163054 c.12A>T p.P4P Chr.3: 120750564-120750564 on Build GRCh38
311678 c.158A>G p.K53R Chr.3: 120750710-120750710 on Build GRCh38
6676680 c.34G>A p.A12T Chr.3: 120750586-120750586 on Build GRCh38
3800938 c.47G>A p.R16Q Chr.3: 120750599-120750599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763096529 Chr.3: 120750671-120750671 on Build GRCh38
rs571206282 Chr.3: 120750515-120750515 on Build GRCh38
rs200639912 Chr.3: 120750508-120750508 on Build GRCh38
rs925122030 Chr.3: 120750568-120750568 on Build GRCh38
rs745598546 Chr.3: 120750537-120750537 on Build GRCh38
rs573424254 Chr.3: 120750634-120750634 on Build GRCh38
rs553760300 Chr.3: 120750616-120750616 on Build GRCh38
rs375390678 Chr.3: 120750619-120750619 on Build GRCh38
rs764223222 Chr.3: 120750681-120750681 on Build GRCh38
rs186444035 Chr.3: 120750575-120750575 on Build GRCh38
rs943707697 Chr.3: 120750600-120750600 on Build GRCh38
rs370873762 Chr.3: 120750536-120750536 on Build GRCh38
rs751623213 Chr.3: 120750685-120750685 on Build GRCh38
rs757356626 Chr.3: 120750606-120750606 on Build GRCh38
rs201779469 Chr.3: 120750620-120750620 on Build GRCh38
rs778517283 Chr.3: 120750583-120750583 on Build GRCh38
rs371166324 Chr.3: 120750573-120750573 on Build GRCh38
rs980528398 Chr.3: 120750663-120750663 on Build GRCh38
rs748156939 Chr.3: 120750628-120750628 on Build GRCh38
rs764597019 Chr.3: 120750580-120750580 on Build GRCh38
rs556947801 Chr.3: 120750546-120750546 on Build GRCh38
rs199539749 Chr.3: 120750542-120750542 on Build GRCh38
rs775517427 Chr.3: 120750648-120750648 on Build GRCh38
rs770606340 Chr.3: 120750506-120750506 on Build GRCh38
rs374032554 Chr.3: 120750525-120750525 on Build GRCh38
rs754462901 Chr.3: 120750505-120750505 on Build GRCh38
rs779528779 Chr.3: 120750615-120750615 on Build GRCh38
rs780824553 Chr.3: 120750521-120750521 on Build GRCh38
rs146072078 Chr.3: 120750703-120750703 on Build GRCh38
rs1024900581 Chr.3: 120750541-120750541 on Build GRCh38
rs748927776 Chr.3: 120750505-120750506 on Build GRCh38
rs765346698 Chr.3: 120750571-120750571 on Build GRCh38
rs150809090 Chr.3: 120750559-120750559 on Build GRCh38
rs755826778 Chr.3: 120750611-120750611 on Build GRCh38
rs772114893 Chr.3: 120750630-120750630 on Build GRCh38
rs878938129 Chr.3: 120750589-120750589 on Build GRCh38
rs368928799 Chr.3: 120750553-120750553 on Build GRCh38
rs770076684 Chr.3: 120750642-120750642 on Build GRCh38
rs775111675 Chr.3: 120750543-120750543 on Build GRCh38
rs762354574 Chr.3: 120750696-120750696 on Build GRCh38
rs200025847 Chr.3: 120750598-120750598 on Build GRCh38
rs374220014 Chr.3: 120750547-120750547 on Build GRCh38
rs748712471 Chr.3: 120750544-120750544 on Build GRCh38
rs968157027 Chr.3: 120750656-120750656 on Build GRCh38
rs758984299 Chr.3: 120750506-120750506 on Build GRCh38
rs767488952 Chr.3: 120750558-120750558 on Build GRCh38
rs774377011 Chr.3: 120750549-120750549 on Build GRCh38
rs757753966 Chr.3: 120750592-120750592 on Build GRCh38
rs759852820 Chr.3: 120750564-120750564 on Build GRCh38
rs751995153 Chr.3: 120750590-120750590 on Build GRCh38

View Full Product Details