Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197059832-197060081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG1
Entrez Gene ID
1739
Gene Name
discs large MAGUK scaffold protein 1
Gene Aliases
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg
UniGene
Hs.292549
Gene Chromosome Location
Chr.3: 197042560-197299272 on Build GRCh38
Transcripts
NM_001098424.1 NM_001204386.1 NM_001204387.1 NM_001204388.1 NM_001290983.1 NM_004087.2 XM_005269289.3 XM_011512502.2 XM_011512503.1 XM_011512505.1 XM_011512506.1 XM_017005800.1 XM_017005801.1 XM_017005802.1 XM_017005803.1 XM_017005804.1 XM_017005805.1 XM_017005806.1 XM_017005807.1 XM_017005808.1 XM_017005809.1 XM_017005810.1 XM_017005811.1 XM_017005812.1 XM_017005813.1 XM_017005814.1 XM_017005815.1 XM_017005816.1 XM_017005817.1 XM_017005818.1 XM_017005819.1 XM_017005820.1 XM_017005821.1 XM_017005822.1 XM_017005823.1 XM_017005824.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1485055 c.2516G>C p.R839T Chr.3: 197059955-197059955 on Build GRCh38
3591916 c.2501C>T p.S834F Chr.3: 197059904-197059904 on Build GRCh38
1485056 c.2450G>C p.R817T Chr.3: 197059955-197059955 on Build GRCh38
4116474 c.2408G>A p.G803D Chr.3: 197059997-197059997 on Build GRCh38
4116473 c.2474G>A p.G825D Chr.3: 197059997-197059997 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560345963 Chr.3: 197059933-197059933 on Build GRCh38
rs752272376 Chr.3: 197060040-197060040 on Build GRCh38
rs930166802 Chr.3: 197060031-197060031 on Build GRCh38
rs773051195 Chr.3: 197060009-197060009 on Build GRCh38
rs563028592 Chr.3: 197060018-197060019 on Build GRCh38
rs752360157 Chr.3: 197059904-197059904 on Build GRCh38
rs953041672 Chr.3: 197059874-197059874 on Build GRCh38
rs776695727 Chr.3: 197059963-197059963 on Build GRCh38
rs377083623 Chr.3: 197059900-197059900 on Build GRCh38
rs531979102 Chr.3: 197060032-197060032 on Build GRCh38
rs759579279 Chr.3: 197060025-197060025 on Build GRCh38
rs191106545 Chr.3: 197059936-197059936 on Build GRCh38
rs546029644 Chr.3: 197059972-197059972 on Build GRCh38
rs762694085 Chr.3: 197060018-197060018 on Build GRCh38
rs747535008 Chr.3: 197059938-197059938 on Build GRCh38
rs760363810 Chr.3: 197060044-197060044 on Build GRCh38
rs751046908 Chr.3: 197059883-197059883 on Build GRCh38
rs775565630 Chr.3: 197059895-197059895 on Build GRCh38
rs768680905 Chr.3: 197059960-197059960 on Build GRCh38
rs746276285 Chr.3: 197059932-197059932 on Build GRCh38
rs769701427 Chr.3: 197059923-197059923 on Build GRCh38
rs992400745 Chr.3: 197060052-197060052 on Build GRCh38
rs779280165 Chr.3: 197059929-197059929 on Build GRCh38
rs754581777 Chr.3: 197059889-197059889 on Build GRCh38
rs950098158 Chr.3: 197060058-197060058 on Build GRCh38
rs564650721 Chr.3: 197060019-197060019 on Build GRCh38
rs748311393 Chr.3: 197059965-197059965 on Build GRCh38
rs767073707 Chr.3: 197060027-197060027 on Build GRCh38

View Full Product Details