Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128246731-128247000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEFSEC
Entrez Gene ID
60678
Gene Name
eukaryotic elongation factor, selenocysteine-tRNA specific
Gene Aliases
EFSEC, SELB
UniGene
Hs.477498
Gene Chromosome Location
Chr.3: 128153456-128426193 on Build GRCh38
Transcripts
NM_021937.4 XM_005247696.2 XM_017006975.1 XM_017006976.1 XM_017006977.1 XM_017006978.1 XM_017006979.1 XM_017006980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651810 c.439delA p.I148fs*1 Chr.3: 128246958-128246958 on Build GRCh38
6651811 c.453A>G p.L151L Chr.3: 128246972-128246972 on Build GRCh38
4491615 c.383C>T p.A128V Chr.3: 128246902-128246902 on Build GRCh38
4113244 c.479T>A p.I160N Chr.3: 128246998-128246998 on Build GRCh38
3059475 c.351C>T p.I117I Chr.3: 128246870-128246870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139234372 Chr.3: 128246778-128246778 on Build GRCh38
rs762501269 Chr.3: 128246891-128246891 on Build GRCh38
rs368182840 Chr.3: 128246798-128246798 on Build GRCh38
rs964589292 Chr.3: 128246776-128246776 on Build GRCh38
rs181759924 Chr.3: 128246973-128246973 on Build GRCh38
rs199938947 Chr.3: 128246835-128246835 on Build GRCh38
rs779169658 Chr.3: 128246945-128246945 on Build GRCh38
rs962983853 Chr.3: 128246844-128246844 on Build GRCh38
rs773114823 Chr.3: 128246830-128246830 on Build GRCh38
rs771550760 Chr.3: 128246792-128246792 on Build GRCh38
rs759054146 Chr.3: 128246871-128246871 on Build GRCh38
rs777862851 Chr.3: 128246962-128246962 on Build GRCh38
rs766788552 Chr.3: 128246918-128246918 on Build GRCh38
rs748259783 Chr.3: 128246824-128246824 on Build GRCh38
rs374889998 Chr.3: 128246903-128246903 on Build GRCh38
rs368860399 Chr.3: 128246946-128246946 on Build GRCh38
rs983579201 Chr.3: 128246928-128246928 on Build GRCh38
rs775338779 Chr.3: 128246948-128246948 on Build GRCh38
rs763718121 Chr.3: 128246897-128246897 on Build GRCh38
rs201961790 Chr.3: 128246840-128246840 on Build GRCh38
rs749402264 Chr.3: 128246799-128246799 on Build GRCh38
rs779065959 Chr.3: 128246816-128246816 on Build GRCh38
rs754213200 Chr.3: 128246921-128246921 on Build GRCh38
rs768932776 Chr.3: 128246800-128246800 on Build GRCh38
rs772182707 Chr.3: 128246825-128246825 on Build GRCh38
rs764937825 Chr.3: 128246876-128246876 on Build GRCh38
rs751013917 Chr.3: 128246788-128246788 on Build GRCh38
rs139131272 Chr.3: 128246924-128246924 on Build GRCh38
rs776489736 Chr.3: 128246863-128246863 on Build GRCh38
rs781755214 Chr.3: 128246787-128246787 on Build GRCh38
rs1036045985 Chr.3: 128246777-128246777 on Build GRCh38
rs150775123 Chr.3: 128246889-128246889 on Build GRCh38
rs775044953 Chr.3: 128246883-128246883 on Build GRCh38
rs569634798 Chr.3: 128246911-128246911 on Build GRCh38
rs746972285 Chr.3: 128246838-128246838 on Build GRCh38
rs746884649 Chr.3: 128246967-128246967 on Build GRCh38

View Full Product Details