Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93750377-93750604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf54
Entrez Gene ID
28970
Gene Name
chromosome 11 open reading frame 54
Gene Aliases
PTD012, PTOD012
UniGene
Hs.8360
Gene Chromosome Location
Chr.11: 93741594-93765323 on Build GRCh38
Transcripts
NM_001286067.1 NM_001286068.1 NM_001286069.1 NM_001286070.1 NM_001286071.1 NM_014039.3 XM_006718824.1 XM_011542781.2 XM_011542782.2 XM_017017613.1 XM_017017614.1 XM_017017615.1 XM_017017616.1 XM_017017617.1 XM_017017618.1 XM_017017619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4201829 c.98T>C p.V33A Chr.11: 93750388-93750388 on Build GRCh38
2224752 c.130G>A p.E44K Chr.11: 93750420-93750420 on Build GRCh38
6838084 c.109G>C p.D37H Chr.11: 93750399-93750399 on Build GRCh38
4201830 c.130G>A p.E44K Chr.11: 93750420-93750420 on Build GRCh38
2224751 c.98T>C p.V33A Chr.11: 93750388-93750388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897794296 Chr.11: 93750412-93750412 on Build GRCh38
rs921996347 Chr.11: 93750521-93750521 on Build GRCh38
rs547581771 Chr.11: 93750415-93750415 on Build GRCh38
rs944821115 Chr.11: 93750401-93750401 on Build GRCh38
rs759251134 Chr.11: 93750461-93750461 on Build GRCh38
rs879192610 Chr.11: 93750464-93750464 on Build GRCh38
rs755304988 Chr.11: 93750406-93750406 on Build GRCh38
rs1044248064 Chr.11: 93750570-93750570 on Build GRCh38
rs748388850 Chr.11: 93750420-93750420 on Build GRCh38
rs772209663 Chr.11: 93750430-93750430 on Build GRCh38
rs747000667 Chr.11: 93750438-93750438 on Build GRCh38
rs773249677 Chr.11: 93750431-93750431 on Build GRCh38
rs769570473 Chr.11: 93750477-93750477 on Build GRCh38
rs770880685 Chr.11: 93750455-93750455 on Build GRCh38
rs368996926 Chr.11: 93750457-93750457 on Build GRCh38
rs946906148 Chr.11: 93750530-93750530 on Build GRCh38

View Full Product Details