Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6887527-6887774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMTA1
Entrez Gene ID
23261
Gene Name
calmodulin binding transcription activator 1
Gene Aliases
CANPMR
UniGene
Hs.397705
Gene Chromosome Location
Chr.1: 6785324-7769706 on Build GRCh38
Transcripts
NM_001242701.1 NM_015215.3 NR_038934.1 XM_011541083.2 XM_011541084.2 XM_011541085.2 XM_011541086.2 XM_011541087.2 XM_011541088.2 XM_011541089.2 XM_011541090.2 XM_011541091.2 XM_011541092.2 XM_017000773.1 XM_017000774.1 XM_017000775.1 XM_017000776.1 XM_017000777.1 XM_017000778.1 XM_017000779.1 XM_017000780.1

SNP Detail

SNP ID Chromosome Location
rs190668117 Chr.1: 6887617-6887617 on Build GRCh38
rs889959625 Chr.1: 6887622-6887622 on Build GRCh38
rs371149140 Chr.1: 6887600-6887600 on Build GRCh38
rs978919736 Chr.1: 6887706-6887706 on Build GRCh38
rs972489702 Chr.1: 6887616-6887616 on Build GRCh38
rs978560448 Chr.1: 6887740-6887740 on Build GRCh38
rs1022857046 Chr.1: 6887707-6887707 on Build GRCh38
rs765926770 Chr.1: 6887641-6887641 on Build GRCh38
rs112339766 Chr.1: 6887618-6887618 on Build GRCh38
rs1049917758 Chr.1: 6887620-6887620 on Build GRCh38
rs988480904 Chr.1: 6887600-6887600 on Build GRCh38
rs941492442 Chr.1: 6887627-6887627 on Build GRCh38
rs1007383994 Chr.1: 6887558-6887558 on Build GRCh38
rs765307011 Chr.1: 6887680-6887680 on Build GRCh38
rs776660122 Chr.1: 6887643-6887643 on Build GRCh38
rs186621821 Chr.1: 6887614-6887614 on Build GRCh38
rs776564748 Chr.1: 6887638-6887638 on Build GRCh38
rs3810982 Chr.1: 6887657-6887657 on Build GRCh38
rs181146054 Chr.1: 6887580-6887580 on Build GRCh38
rs934906243 Chr.1: 6887729-6887729 on Build GRCh38
rs897209162 Chr.1: 6887654-6887654 on Build GRCh38
rs1035010156 Chr.1: 6887667-6887667 on Build GRCh38
rs926200557 Chr.1: 6887717-6887717 on Build GRCh38
rs992937245 Chr.1: 6887656-6887656 on Build GRCh38
rs764758251 Chr.1: 6887754-6887754 on Build GRCh38
rs934331715 Chr.1: 6887593-6887593 on Build GRCh38

View Full Product Details