Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93075753-93076020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP26A1
Entrez Gene ID
1592
Gene Name
cytochrome P450 family 26 subfamily A member 1
Gene Aliases
CP26, CYP26, P450RAI, P450RAI1
UniGene
Hs.150595
Gene Chromosome Location
Chr.10: 93073475-93077884 on Build GRCh38
Transcripts
NM_000783.3 NM_057157.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4513839 c.745C>T p.L249F Chr.10: 93075913-93075913 on Build GRCh38
5473057 c.792+8G>T p.? Chr.10: 93075968-93075968 on Build GRCh38
3395990 c.979C>T p.R327* Chr.10: 93075940-93075940 on Build GRCh38
6551159 c.987G>T p.E329D Chr.10: 93075948-93075948 on Build GRCh38
921538 c.786G>T p.K262N Chr.10: 93075954-93075954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41290186 Chr.10: 93075818-93075818 on Build GRCh38
rs1047023575 Chr.10: 93075799-93075799 on Build GRCh38
rs369951759 Chr.10: 93075943-93075943 on Build GRCh38
rs769135070 Chr.10: 93075974-93075974 on Build GRCh38
rs375356227 Chr.10: 93075784-93075784 on Build GRCh38
rs142962735 Chr.10: 93075875-93075875 on Build GRCh38
rs763307370 Chr.10: 93075884-93075884 on Build GRCh38
rs777309274 Chr.10: 93075933-93075933 on Build GRCh38
rs754813478 Chr.10: 93075902-93075902 on Build GRCh38
rs868634165 Chr.10: 93075862-93075862 on Build GRCh38
rs772703300 Chr.10: 93075838-93075838 on Build GRCh38
rs758532215 Chr.10: 93075819-93075821 on Build GRCh38
rs777669352 Chr.10: 93075869-93075870 on Build GRCh38
rs1035377847 Chr.10: 93075839-93075839 on Build GRCh38
rs762226337 Chr.10: 93075871-93075871 on Build GRCh38
rs777202473 Chr.10: 93075866-93075866 on Build GRCh38
rs74435030 Chr.10: 93075874-93075874 on Build GRCh38
rs374621068 Chr.10: 93075968-93075968 on Build GRCh38
rs1014855726 Chr.10: 93075940-93075940 on Build GRCh38
rs751784359 Chr.10: 93075901-93075901 on Build GRCh38
rs776849925 Chr.10: 93075979-93075979 on Build GRCh38
rs769349916 Chr.10: 93075832-93075832 on Build GRCh38
rs778556527 Chr.10: 93075970-93075970 on Build GRCh38
rs1030619383 Chr.10: 93075981-93075981 on Build GRCh38
rs892606997 Chr.10: 93075785-93075785 on Build GRCh38
rs781214654 Chr.10: 93075828-93075828 on Build GRCh38
rs766739833 Chr.10: 93075891-93075891 on Build GRCh38
rs547969483 Chr.10: 93075991-93075991 on Build GRCh38
rs780782441 Chr.10: 93075915-93075915 on Build GRCh38
rs758683977 Chr.10: 93075812-93075812 on Build GRCh38
rs755991400 Chr.10: 93075931-93075931 on Build GRCh38
rs1006070001 Chr.10: 93075938-93075938 on Build GRCh38
rs745447535 Chr.10: 93075973-93075973 on Build GRCh38
rs565440327 Chr.10: 93075852-93075852 on Build GRCh38
rs369345955 Chr.10: 93075813-93075813 on Build GRCh38
rs747975009 Chr.10: 93075831-93075831 on Build GRCh38
rs773366572 Chr.10: 93075846-93075846 on Build GRCh38
rs750744997 Chr.10: 93075798-93075798 on Build GRCh38
rs762432003 Chr.10: 93075982-93075982 on Build GRCh38
rs752745616 Chr.10: 93075925-93075925 on Build GRCh38
rs773819065 Chr.10: 93075994-93075994 on Build GRCh38
rs748998239 Chr.10: 93075844-93075844 on Build GRCh38
rs1016025294 Chr.10: 93075803-93075803 on Build GRCh38
rs373196632 Chr.10: 93075815-93075815 on Build GRCh38
rs768851695 Chr.10: 93075863-93075863 on Build GRCh38
rs746746199 Chr.10: 93075958-93075959 on Build GRCh38
rs754691610 Chr.10: 93075825-93075825 on Build GRCh38

View Full Product Details