Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24507295-24507568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMA1
Entrez Gene ID
1215
Gene Name
chymase 1
Gene Aliases
CYH, MCT1, chymase
UniGene
Hs.135626
Gene Chromosome Location
Chr.14: 24505355-24508265 on Build GRCh38
Transcripts
NM_001308083.1 NM_001836.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6223756 c.209G>T p.R70M Chr.14: 24507356-24507356 on Build GRCh38
6630897 c.88C>A p.P30T Chr.14: 24507477-24507477 on Build GRCh38
6075652 c.105C>A p.Y35* Chr.14: 24507460-24507460 on Build GRCh38
363084 c.97C>A p.R33S Chr.14: 24507468-24507468 on Build GRCh38
6630894 c.135C>T p.N45N Chr.14: 24507430-24507430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773464372 Chr.14: 24507490-24507490 on Build GRCh38
rs13306251 Chr.14: 24507377-24507377 on Build GRCh38
rs140753864 Chr.14: 24507495-24507495 on Build GRCh38
rs748332591 Chr.14: 24507433-24507433 on Build GRCh38
rs755577285 Chr.14: 24507539-24507539 on Build GRCh38
rs748509171 Chr.14: 24507485-24507485 on Build GRCh38
rs200210391 Chr.14: 24507388-24507388 on Build GRCh38
rs942878411 Chr.14: 24507520-24507520 on Build GRCh38
rs755008668 Chr.14: 24507370-24507371 on Build GRCh38
rs755797763 Chr.14: 24507336-24507336 on Build GRCh38
rs377088231 Chr.14: 24507437-24507437 on Build GRCh38
rs777466346 Chr.14: 24507399-24507399 on Build GRCh38
rs995567512 Chr.14: 24507464-24507464 on Build GRCh38
rs556132795 Chr.14: 24507345-24507345 on Build GRCh38
rs5246 Chr.14: 24507429-24507429 on Build GRCh38
rs765236468 Chr.14: 24507376-24507376 on Build GRCh38
rs748992941 Chr.14: 24507490-24507491 on Build GRCh38
rs539562095 Chr.14: 24507359-24507359 on Build GRCh38
rs138432864 Chr.14: 24507468-24507468 on Build GRCh38
rs747437697 Chr.14: 24507426-24507426 on Build GRCh38
rs754878710 Chr.14: 24507325-24507325 on Build GRCh38
rs376957072 Chr.14: 24507496-24507496 on Build GRCh38
rs752443584 Chr.14: 24507335-24507335 on Build GRCh38
rs758059013 Chr.14: 24507527-24507527 on Build GRCh38
rs753433709 Chr.14: 24507404-24507404 on Build GRCh38
rs549266517 Chr.14: 24507456-24507456 on Build GRCh38
rs1023375757 Chr.14: 24507424-24507424 on Build GRCh38
rs748063446 Chr.14: 24507373-24507374 on Build GRCh38
rs542619483 Chr.14: 24507399-24507399 on Build GRCh38
rs201020585 Chr.14: 24507467-24507467 on Build GRCh38
rs552670864 Chr.14: 24507488-24507488 on Build GRCh38
rs202195650 Chr.14: 24507393-24507393 on Build GRCh38
rs987017084 Chr.14: 24507328-24507328 on Build GRCh38
rs763885364 Chr.14: 24507457-24507457 on Build GRCh38
rs373630806 Chr.14: 24507342-24507342 on Build GRCh38
rs761088628 Chr.14: 24507363-24507363 on Build GRCh38
rs370555372 Chr.14: 24507338-24507338 on Build GRCh38
rs5247 Chr.14: 24507368-24507368 on Build GRCh38
rs780997300 Chr.14: 24507371-24507372 on Build GRCh38
rs1001813145 Chr.14: 24507430-24507430 on Build GRCh38
rs183530742 Chr.14: 24507499-24507499 on Build GRCh38
rs755484632 Chr.14: 24507474-24507474 on Build GRCh38
rs753617059 Chr.14: 24507459-24507459 on Build GRCh38
rs763060029 Chr.14: 24507452-24507452 on Build GRCh38
rs746792590 Chr.14: 24507366-24507367 on Build GRCh38
rs762021583 Chr.14: 24507372-24507372 on Build GRCh38
rs776027325 Chr.14: 24507362-24507362 on Build GRCh38
rs931343730 Chr.14: 24507360-24507360 on Build GRCh38
rs775687715 Chr.14: 24507374-24507374 on Build GRCh38
rs776982970 Chr.14: 24507369-24507369 on Build GRCh38
rs760625006 Chr.14: 24507497-24507497 on Build GRCh38
rs770690012 Chr.14: 24507525-24507525 on Build GRCh38
rs778307845 Chr.14: 24507411-24507411 on Build GRCh38
rs150310098 Chr.14: 24507355-24507355 on Build GRCh38
rs372192946 Chr.14: 24507392-24507392 on Build GRCh38
rs368187838 Chr.14: 24507441-24507441 on Build GRCh38
rs748370921 Chr.14: 24507522-24507522 on Build GRCh38
rs766000872 Chr.14: 24507536-24507536 on Build GRCh38
rs745343471 Chr.14: 24507413-24507413 on Build GRCh38
rs770339788 Chr.14: 24507343-24507343 on Build GRCh38
rs145340056 Chr.14: 24507466-24507466 on Build GRCh38
rs142904150 Chr.14: 24507422-24507422 on Build GRCh38
rs772739296 Chr.14: 24507479-24507479 on Build GRCh38
rs767494660 Chr.14: 24507455-24507455 on Build GRCh38
rs932232367 Chr.14: 24507481-24507481 on Build GRCh38
rs78253044 Chr.14: 24507334-24507334 on Build GRCh38
rs200189048 Chr.14: 24507410-24507410 on Build GRCh38
rs751066117 Chr.14: 24507537-24507537 on Build GRCh38
rs761645413 Chr.14: 24507518-24507518 on Build GRCh38
rs778416592 Chr.14: 24507354-24507354 on Build GRCh38
rs896348903 Chr.14: 24507403-24507403 on Build GRCh38
rs781433490 Chr.14: 24507477-24507477 on Build GRCh38
rs13306254 Chr.14: 24507451-24507451 on Build GRCh38
rs776902640 Chr.14: 24507502-24507502 on Build GRCh38

View Full Product Details