Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132571224-132571477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBRSL1
Entrez Gene ID
57666
Gene Name
fibrosin like 1
Gene Aliases
-
UniGene
Hs.411138
Gene Chromosome Location
Chr.12: 132490440-132585188 on Build GRCh38
Transcripts
NM_001142641.1 XM_005266171.3 XM_005266173.3 XM_005266175.3 XM_005266176.3 XM_005266177.3 XM_011534803.2 XM_011534804.2 XM_011534805.2 XM_011534806.2 XM_011534807.2 XM_011534808.2 XM_011534809.2 XM_011534810.2 XM_011534812.2 XM_011534816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1206757 c.172C>T p.R58W Chr.12: 132571399-132571399 on Build GRCh38
6244589 c.168G>T p.L56L Chr.12: 132571395-132571395 on Build GRCh38
1206759 c.136C>T p.R46W Chr.12: 132571363-132571363 on Build GRCh38
3687867 c.195G>A p.Q65Q Chr.12: 132571422-132571422 on Build GRCh38
7291516 c.1278C>T p.G426G Chr.12: 132571371-132571371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754612789 Chr.12: 132571459-132571459 on Build GRCh38
rs752732640 Chr.12: 132571423-132571446 on Build GRCh38
rs778402555 Chr.12: 132571276-132571276 on Build GRCh38
rs539638805 Chr.12: 132571262-132571262 on Build GRCh38
rs752091289 Chr.12: 132571276-132571277 on Build GRCh38
rs140368393 Chr.12: 132571246-132571246 on Build GRCh38
rs1025553901 Chr.12: 132571364-132571364 on Build GRCh38
rs774053548 Chr.12: 132571282-132571283 on Build GRCh38
rs776793201 Chr.12: 132571296-132571296 on Build GRCh38
rs781265803 Chr.12: 132571417-132571428 on Build GRCh38
rs1016487005 Chr.12: 132571329-132571329 on Build GRCh38
rs569489036 Chr.12: 132571356-132571356 on Build GRCh38
rs906191712 Chr.12: 132571337-132571337 on Build GRCh38
rs770873432 Chr.12: 132571292-132571292 on Build GRCh38
rs746016516 Chr.12: 132571340-132571340 on Build GRCh38
rs1046126580 Chr.12: 132571315-132571315 on Build GRCh38
rs1026666610 Chr.12: 132571440-132571440 on Build GRCh38
rs1031339536 Chr.12: 132571404-132571404 on Build GRCh38
rs955865059 Chr.12: 132571441-132571441 on Build GRCh38
rs1047762821 Chr.12: 132571286-132571286 on Build GRCh38
rs914136236 Chr.12: 132571311-132571311 on Build GRCh38
rs765761087 Chr.12: 132571439-132571439 on Build GRCh38
rs775751741 Chr.12: 132571363-132571363 on Build GRCh38
rs755501464 Chr.12: 132571283-132571284 on Build GRCh38
rs1006487953 Chr.12: 132571312-132571312 on Build GRCh38
rs760701483 Chr.12: 132571399-132571399 on Build GRCh38
rs772632650 Chr.12: 132571270-132571270 on Build GRCh38
rs376821153 Chr.12: 132571264-132571264 on Build GRCh38
rs368308315 Chr.12: 132571385-132571385 on Build GRCh38
rs971207396 Chr.12: 132571367-132571367 on Build GRCh38
rs555972870 Chr.12: 132571281-132571281 on Build GRCh38
rs753403621 Chr.12: 132571447-132571447 on Build GRCh38
rs772995564 Chr.12: 132571386-132571386 on Build GRCh38
rs370956654 Chr.12: 132571265-132571265 on Build GRCh38
rs375193157 Chr.12: 132571371-132571371 on Build GRCh38
rs895336919 Chr.12: 132571339-132571339 on Build GRCh38
rs982721149 Chr.12: 132571247-132571247 on Build GRCh38

View Full Product Details