Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 48963605-48963840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSHR
Entrez Gene ID
2492
Gene Name
follicle stimulating hormone receptor
Gene Aliases
FSHR1, FSHRO, LGR1, ODG1
UniGene
Hs.1428
Gene Chromosome Location
Chr.2: 48953161-49154527 on Build GRCh38
Transcripts
NM_000145.3 NM_181446.2 XM_011532733.2 XM_011532734.2 XM_011532735.2 XM_011532736.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3407888 c.1213G>A p.A405T Chr.2: 48963608-48963608 on Build GRCh38
4855226 c.1125G>A p.L375L Chr.2: 48963696-48963696 on Build GRCh38
6667490 c.1175A>G p.Y392C Chr.2: 48963646-48963646 on Build GRCh38
721653 c.1018G>A p.E340K Chr.2: 48963803-48963803 on Build GRCh38
4439600 c.1196T>G p.F399C Chr.2: 48963625-48963625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777861273 Chr.2: 48963719-48963719 on Build GRCh38
rs772756688 Chr.2: 48963791-48963791 on Build GRCh38
rs559544903 Chr.2: 48963797-48963797 on Build GRCh38
rs776759086 Chr.2: 48963742-48963742 on Build GRCh38
rs777679833 Chr.2: 48963813-48963813 on Build GRCh38
rs779466348 Chr.2: 48963773-48963773 on Build GRCh38
rs139226976 Chr.2: 48963799-48963799 on Build GRCh38
rs540845861 Chr.2: 48963633-48963633 on Build GRCh38
rs386833510 Chr.2: 48963778-48963778 on Build GRCh38
rs903235388 Chr.2: 48963654-48963654 on Build GRCh38
rs761504646 Chr.2: 48963656-48963656 on Build GRCh38
rs755003829 Chr.2: 48963696-48963696 on Build GRCh38
rs759046473 Chr.2: 48963667-48963667 on Build GRCh38
rs377596075 Chr.2: 48963676-48963676 on Build GRCh38
rs751776676 Chr.2: 48963650-48963650 on Build GRCh38
rs763302784 Chr.2: 48963806-48963806 on Build GRCh38
rs767103311 Chr.2: 48963664-48963664 on Build GRCh38
rs769247170 Chr.2: 48963780-48963780 on Build GRCh38
rs924691070 Chr.2: 48963798-48963798 on Build GRCh38
rs758606613 Chr.2: 48963707-48963707 on Build GRCh38
rs778825740 Chr.2: 48963701-48963701 on Build GRCh38
rs753820515 Chr.2: 48963692-48963692 on Build GRCh38
rs991651509 Chr.2: 48963734-48963734 on Build GRCh38
rs747050487 Chr.2: 48963733-48963733 on Build GRCh38
rs770983180 Chr.2: 48963738-48963738 on Build GRCh38
rs747914737 Chr.2: 48963703-48963703 on Build GRCh38
rs760439697 Chr.2: 48963792-48963792 on Build GRCh38
rs752469633 Chr.2: 48963805-48963805 on Build GRCh38
rs1033006179 Chr.2: 48963709-48963709 on Build GRCh38
rs763262835 Chr.2: 48963776-48963776 on Build GRCh38
rs148784693 Chr.2: 48963812-48963812 on Build GRCh38
rs746345111 Chr.2: 48963768-48963768 on Build GRCh38
rs917432692 Chr.2: 48963749-48963749 on Build GRCh38
rs200532372 Chr.2: 48963648-48963648 on Build GRCh38
rs373696730 Chr.2: 48963672-48963672 on Build GRCh38
rs769202588 Chr.2: 48963811-48963811 on Build GRCh38
rs775786667 Chr.2: 48963774-48963774 on Build GRCh38

View Full Product Details