Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43492812-43493079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX8
Entrez Gene ID
1659
Gene Name
DEAH-box helicase 8
Gene Aliases
DDX8, HRH1, PRP22, PRPF22
UniGene
Hs.463105
Gene Chromosome Location
Chr.17: 43483865-43544787 on Build GRCh38
Transcripts
NM_001302623.1 NM_001322216.1 NM_001322217.1 NM_001322218.1 NM_001322219.1 NM_001322220.1 NM_001322221.1 NM_004941.2 NR_136225.1 NR_136226.1 NR_136227.1 NR_136228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3819708 c.812A>T p.K271I Chr.17: 43492989-43492989 on Build GRCh38
3819707 c.772C>G p.P258A Chr.17: 43492949-43492949 on Build GRCh38
3370744 c.638G>T p.R213L Chr.17: 43492815-43492815 on Build GRCh38
1479619 c.765C>A p.D255E Chr.17: 43492942-43492942 on Build GRCh38
4066791 c.767G>A p.R256H Chr.17: 43492944-43492944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199963654 Chr.17: 43492907-43492907 on Build GRCh38
rs750914324 Chr.17: 43493056-43493056 on Build GRCh38
rs758701682 Chr.17: 43492840-43492840 on Build GRCh38
rs751851397 Chr.17: 43492923-43492923 on Build GRCh38
rs144158744 Chr.17: 43492991-43492991 on Build GRCh38
rs767342717 Chr.17: 43492895-43492895 on Build GRCh38
rs775790218 Chr.17: 43492875-43492875 on Build GRCh38
rs750870534 Chr.17: 43492955-43492955 on Build GRCh38
rs752465390 Chr.17: 43493048-43493048 on Build GRCh38
rs748941253 Chr.17: 43492839-43492839 on Build GRCh38
rs200430936 Chr.17: 43492943-43492943 on Build GRCh38
rs754652647 Chr.17: 43492917-43492917 on Build GRCh38
rs778391111 Chr.17: 43493043-43493043 on Build GRCh38
rs774046464 Chr.17: 43492887-43492887 on Build GRCh38
rs768523273 Chr.17: 43492886-43492886 on Build GRCh38
rs761708865 Chr.17: 43492894-43492894 on Build GRCh38
rs139381870 Chr.17: 43492939-43492939 on Build GRCh38
rs750440731 Chr.17: 43492899-43492899 on Build GRCh38
rs753437233 Chr.17: 43492913-43492913 on Build GRCh38
rs984668514 Chr.17: 43492856-43492856 on Build GRCh38
rs753310286 Chr.17: 43493033-43493033 on Build GRCh38
rs764231495 Chr.17: 43492880-43492880 on Build GRCh38
rs756462291 Chr.17: 43492989-43492989 on Build GRCh38
rs770068955 Chr.17: 43492871-43492871 on Build GRCh38
rs2279191 Chr.17: 43493059-43493059 on Build GRCh38
rs752031496 Chr.17: 43492902-43492902 on Build GRCh38
rs778017608 Chr.17: 43492844-43492844 on Build GRCh38
rs1034134021 Chr.17: 43493022-43493022 on Build GRCh38
rs760625283 Chr.17: 43492998-43492998 on Build GRCh38
rs747507470 Chr.17: 43492845-43492845 on Build GRCh38
rs886296612 Chr.17: 43492947-43492947 on Build GRCh38
rs879524424 Chr.17: 43492889-43492889 on Build GRCh38
rs763284184 Chr.17: 43492881-43492881 on Build GRCh38
rs761508884 Chr.17: 43492968-43492968 on Build GRCh38
rs749482546 Chr.17: 43492944-43492944 on Build GRCh38
rs375354059 Chr.17: 43492868-43492868 on Build GRCh38
rs765833011 Chr.17: 43493028-43493028 on Build GRCh38
rs777247620 Chr.17: 43492854-43492854 on Build GRCh38
rs746384849 Chr.17: 43492926-43492926 on Build GRCh38
rs565443799 Chr.17: 43492954-43492954 on Build GRCh38
rs778663785 Chr.17: 43492921-43492921 on Build GRCh38
rs982093471 Chr.17: 43492999-43492999 on Build GRCh38
rs765884874 Chr.17: 43492911-43492911 on Build GRCh38
rs556024804 Chr.17: 43493053-43493053 on Build GRCh38
rs200639945 Chr.17: 43493036-43493036 on Build GRCh38
rs762089253 Chr.17: 43492993-43492994 on Build GRCh38
rs774692689 Chr.17: 43492966-43492966 on Build GRCh38
rs776820139 Chr.17: 43492923-43492925 on Build GRCh38
rs146727331 Chr.17: 43492925-43492925 on Build GRCh38
rs1024195130 Chr.17: 43492965-43492965 on Build GRCh38
rs757500736 Chr.17: 43493052-43493052 on Build GRCh38
rs771903921 Chr.17: 43492975-43492975 on Build GRCh38

View Full Product Details