Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 83673335-83673608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAPLN1
Entrez Gene ID
1404
Gene Name
hyaluronan and proteoglycan link protein 1
Gene Aliases
CRT1, CRTL1
UniGene
Hs.2799
Gene Chromosome Location
Chr.5: 83638198-83721210 on Build GRCh38
Transcripts
NM_001884.3 XM_011543168.2 XM_017009051.1 XM_017009052.1 XM_017009053.1 XM_017009054.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4497576 c.49C>T p.H17Y Chr.5: 83673475-83673475 on Build GRCh38
6230235 c.38G>A p.C13Y Chr.5: 83673486-83673486 on Build GRCh38
294022 c.72G>A p.L24L Chr.5: 83673452-83673452 on Build GRCh38
373195 c.100G>A p.A34T Chr.5: 83673424-83673424 on Build GRCh38
3139835 c.25C>T p.L9L Chr.5: 83673499-83673499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373107913 Chr.5: 83673554-83673554 on Build GRCh38
rs1027506788 Chr.5: 83673458-83673458 on Build GRCh38
rs1051256289 Chr.5: 83673366-83673366 on Build GRCh38
rs760128581 Chr.5: 83673426-83673426 on Build GRCh38
rs745915783 Chr.5: 83673377-83673377 on Build GRCh38
rs780377189 Chr.5: 83673384-83673384 on Build GRCh38
rs375787364 Chr.5: 83673464-83673464 on Build GRCh38
rs553434815 Chr.5: 83673364-83673364 on Build GRCh38
rs753357240 Chr.5: 83673489-83673489 on Build GRCh38
rs939943040 Chr.5: 83673513-83673513 on Build GRCh38
rs762296938 Chr.5: 83673408-83673408 on Build GRCh38
rs768747470 Chr.5: 83673512-83673512 on Build GRCh38
rs139340085 Chr.5: 83673439-83673439 on Build GRCh38
rs774576911 Chr.5: 83673400-83673400 on Build GRCh38
rs144082755 Chr.5: 83673448-83673448 on Build GRCh38
rs769000140 Chr.5: 83673392-83673392 on Build GRCh38
rs781337723 Chr.5: 83673566-83673566 on Build GRCh38
rs375728164 Chr.5: 83673569-83673569 on Build GRCh38
rs761532834 Chr.5: 83673504-83673506 on Build GRCh38
rs773649016 Chr.5: 83673532-83673532 on Build GRCh38
rs772547488 Chr.5: 83673414-83673414 on Build GRCh38
rs772448036 Chr.5: 83673516-83673516 on Build GRCh38
rs150120434 Chr.5: 83673515-83673515 on Build GRCh38
rs975143594 Chr.5: 83673435-83673435 on Build GRCh38
rs202185751 Chr.5: 83673452-83673452 on Build GRCh38
rs761173044 Chr.5: 83673539-83673539 on Build GRCh38
rs564699077 Chr.5: 83673585-83673585 on Build GRCh38
rs1048746373 Chr.5: 83673463-83673463 on Build GRCh38
rs749453113 Chr.5: 83673491-83673491 on Build GRCh38
rs773738875 Chr.5: 83673422-83673422 on Build GRCh38
rs763998716 Chr.5: 83673564-83673564 on Build GRCh38
rs779245334 Chr.5: 83673514-83673514 on Build GRCh38
rs762610166 Chr.5: 83673556-83673556 on Build GRCh38
rs755127239 Chr.5: 83673574-83673574 on Build GRCh38
rs764995770 Chr.5: 83673455-83673455 on Build GRCh38
rs758250030 Chr.5: 83673456-83673456 on Build GRCh38
rs931144253 Chr.5: 83673453-83673453 on Build GRCh38
rs545911942 Chr.5: 83673553-83673553 on Build GRCh38
rs751573843 Chr.5: 83673473-83673473 on Build GRCh38
rs770029046 Chr.5: 83673379-83673379 on Build GRCh38
rs138357388 Chr.5: 83673542-83673542 on Build GRCh38
rs756207508 Chr.5: 83673475-83673475 on Build GRCh38
rs112149696 Chr.5: 83673411-83673411 on Build GRCh38
rs751479189 Chr.5: 83673565-83673565 on Build GRCh38
rs780289242 Chr.5: 83673476-83673476 on Build GRCh38
rs771491900 Chr.5: 83673541-83673541 on Build GRCh38
rs753842393 Chr.5: 83673567-83673567 on Build GRCh38
rs73769334 Chr.5: 83673385-83673385 on Build GRCh38

View Full Product Details