Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68676276-68676533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH3
Entrez Gene ID
1001
Gene Name
cadherin 3
Gene Aliases
CDHP, HJMD, PCAD
UniGene
Hs.191842
Gene Chromosome Location
Chr.16: 68644820-68727573 on Build GRCh38
Transcripts
NM_001317195.1 NM_001317196.1 NM_001793.5 XM_011522800.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4569942 c.218T>A p.F73Y Chr.16: 68676442-68676442 on Build GRCh38
2996615 c.245A>G p.Q82R Chr.16: 68676469-68676469 on Build GRCh38
1179085 c.235G>A p.E79K Chr.16: 68676459-68676459 on Build GRCh38
4569941 c.218T>A p.F73Y Chr.16: 68676442-68676442 on Build GRCh38
1679218 c.226C>T p.R76W Chr.16: 68676450-68676450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139427769 Chr.16: 68676362-68676362 on Build GRCh38
rs755650735 Chr.16: 68676386-68676386 on Build GRCh38
rs1036002643 Chr.16: 68676390-68676390 on Build GRCh38
rs767009501 Chr.16: 68676446-68676446 on Build GRCh38
rs780496426 Chr.16: 68676314-68676315 on Build GRCh38
rs201456844 Chr.16: 68676457-68676457 on Build GRCh38
rs376051800 Chr.16: 68676476-68676476 on Build GRCh38
rs779463050 Chr.16: 68676412-68676412 on Build GRCh38
rs775594020 Chr.16: 68676338-68676338 on Build GRCh38
rs766015706 Chr.16: 68676481-68676481 on Build GRCh38
rs879086126 Chr.16: 68676368-68676368 on Build GRCh38
rs753404769 Chr.16: 68676486-68676486 on Build GRCh38
rs562980302 Chr.16: 68676499-68676502 on Build GRCh38
rs539696131 Chr.16: 68676423-68676423 on Build GRCh38
rs777890333 Chr.16: 68676444-68676444 on Build GRCh38
rs200141594 Chr.16: 68676498-68676498 on Build GRCh38
rs779490510 Chr.16: 68676394-68676394 on Build GRCh38
rs886052228 Chr.16: 68676390-68676390 on Build GRCh38
rs754684451 Chr.16: 68676417-68676418 on Build GRCh38
rs554880661 Chr.16: 68676496-68676496 on Build GRCh38
rs764057460 Chr.16: 68676347-68676347 on Build GRCh38
rs868126624 Chr.16: 68676502-68676502 on Build GRCh38
rs374865506 Chr.16: 68676451-68676451 on Build GRCh38
rs201122541 Chr.16: 68676458-68676458 on Build GRCh38
rs4359428 Chr.16: 68676307-68676307 on Build GRCh38
rs751374394 Chr.16: 68676352-68676352 on Build GRCh38
rs760369421 Chr.16: 68676479-68676479 on Build GRCh38
rs193921054 Chr.16: 68676459-68676459 on Build GRCh38
rs372990007 Chr.16: 68676344-68676344 on Build GRCh38
rs113195974 Chr.16: 68676477-68676477 on Build GRCh38
rs538255856 Chr.16: 68676438-68676438 on Build GRCh38
rs1007376972 Chr.16: 68676505-68676505 on Build GRCh38
rs988960610 Chr.16: 68676414-68676414 on Build GRCh38
rs751934830 Chr.16: 68676376-68676377 on Build GRCh38
rs754454265 Chr.16: 68676487-68676487 on Build GRCh38
rs571492325 Chr.16: 68676506-68676506 on Build GRCh38
rs767087158 Chr.16: 68676370-68676370 on Build GRCh38
rs767301443 Chr.16: 68676471-68676471 on Build GRCh38
rs546610061 Chr.16: 68676450-68676450 on Build GRCh38
rs747517754 Chr.16: 68676375-68676375 on Build GRCh38
rs748477392 Chr.16: 68676417-68676417 on Build GRCh38
rs144998887 Chr.16: 68676377-68676377 on Build GRCh38
rs759681036 Chr.16: 68676454-68676454 on Build GRCh38
rs761515421 Chr.16: 68676463-68676463 on Build GRCh38
rs753323144 Chr.16: 68676397-68676397 on Build GRCh38
rs191945631 Chr.16: 68676402-68676402 on Build GRCh38
rs1056047936 Chr.16: 68676319-68676319 on Build GRCh38
rs747182343 Chr.16: 68676446-68676446 on Build GRCh38
rs375382810 Chr.16: 68676514-68676514 on Build GRCh38

View Full Product Details