Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 64950900-64951127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH11
Entrez Gene ID
1009
Gene Name
cadherin 11
Gene Aliases
CAD11, CDHOB, OB, OSF-4
UniGene
Hs.116471
Gene Chromosome Location
Chr.16: 64943753-65126112 on Build GRCh38
Transcripts
NM_001308392.1 NM_001797.3 XM_005255761.3 XM_005255762.2 XM_005255763.2 XM_011522803.2 XM_017022830.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1645749 c.1663G>T p.A555S Chr.16: 64950998-64950998 on Build GRCh38
5740673 c.1694A>G p.Q565R Chr.16: 64950967-64950967 on Build GRCh38
6988630 c.1663G>A p.A555T Chr.16: 64950998-64950998 on Build GRCh38
1645750 c.1712C>T p.P571L Chr.16: 64950949-64950949 on Build GRCh38
4872932 c.1726G>A p.D576N Chr.16: 64950935-64950935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747064758 Chr.16: 64950997-64950997 on Build GRCh38
rs572136896 Chr.16: 64951082-64951082 on Build GRCh38
rs749905567 Chr.16: 64950939-64950939 on Build GRCh38
rs118105117 Chr.16: 64951062-64951062 on Build GRCh38
rs138180051 Chr.16: 64951009-64951009 on Build GRCh38
rs200486624 Chr.16: 64950999-64950999 on Build GRCh38
rs971498235 Chr.16: 64951046-64951046 on Build GRCh38
rs879077857 Chr.16: 64951008-64951008 on Build GRCh38
rs139147793 Chr.16: 64950933-64950933 on Build GRCh38
rs767438277 Chr.16: 64951013-64951013 on Build GRCh38
rs772565642 Chr.16: 64950993-64950993 on Build GRCh38
rs911983831 Chr.16: 64951030-64951030 on Build GRCh38
rs759807372 Chr.16: 64950921-64950921 on Build GRCh38
rs754745285 Chr.16: 64950976-64950976 on Build GRCh38
rs753510203 Chr.16: 64950963-64950963 on Build GRCh38
rs984796718 Chr.16: 64951073-64951073 on Build GRCh38
rs905742600 Chr.16: 64951087-64951087 on Build GRCh38
rs866744133 Chr.16: 64950928-64950928 on Build GRCh38
rs147033764 Chr.16: 64950995-64950995 on Build GRCh38
rs201534694 Chr.16: 64950959-64950959 on Build GRCh38
rs761749550 Chr.16: 64950927-64950927 on Build GRCh38
rs556854549 Chr.16: 64951012-64951012 on Build GRCh38
rs767357192 Chr.16: 64950930-64950930 on Build GRCh38
rs565044007 Chr.16: 64951061-64951061 on Build GRCh38
rs772999496 Chr.16: 64951029-64951029 on Build GRCh38
rs760705911 Chr.16: 64951039-64951039 on Build GRCh38
rs889460792 Chr.16: 64950972-64950972 on Build GRCh38
rs776863801 Chr.16: 64950920-64950920 on Build GRCh38
rs779408399 Chr.16: 64950977-64950977 on Build GRCh38
rs373589644 Chr.16: 64951002-64951002 on Build GRCh38
rs759737776 Chr.16: 64951005-64951005 on Build GRCh38
rs765982483 Chr.16: 64951042-64951042 on Build GRCh38
rs755516686 Chr.16: 64951047-64951047 on Build GRCh38
rs776736292 Chr.16: 64951004-64951004 on Build GRCh38
rs909332073 Chr.16: 64951091-64951091 on Build GRCh38
rs953517296 Chr.16: 64951018-64951018 on Build GRCh38
rs763850377 Chr.16: 64950922-64950922 on Build GRCh38
rs369835570 Chr.16: 64951048-64951048 on Build GRCh38
rs185190786 Chr.16: 64950991-64950991 on Build GRCh38
rs869205451 Chr.16: 64951062-64951062 on Build GRCh38
rs369832077 Chr.16: 64950941-64950941 on Build GRCh38
rs878906742 Chr.16: 64950924-64950924 on Build GRCh38
rs142957030 Chr.16: 64950953-64950953 on Build GRCh38
rs558670899 Chr.16: 64951074-64951074 on Build GRCh38

View Full Product Details