Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDR
Entrez Gene ID
3791
Gene Name
kinase insert domain receptor
Gene Aliases
CD309, FLK1, VEGFR, VEGFR2
Chromosome Location
Chr.4: 55078259 - 55125595 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002253.2 NP_002244.1 22 - 23 3371 114
GenBank mRNA AB209901.1 - 22 - 23 3356 114
AF035121.1 - 22 - 23 3372 114
AF063658.1 - 22 - 23 3069 114
BC131822.1 - 22 - 23 3131 114
L04947.1 - 22 - 23 3063 114
X61656.1 - 22 - 23 3063 114

View Full Product Details