Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIRT1
Entrez Gene ID
23411
Gene Name
sirtuin 1
Gene Aliases
SIR2, SIR2L1, SIR2alpha
Chromosome Location
Chr.10: 67884669 - 67918390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012238.4 NP_036370.2 2 - 3 599 74
GenBank mRNA AF083106.2 - 2 - 3 599 74
AF235040.1 - 2 - 3 594 74
AK289743.1 - 2 - 3 335 74
AL136741.1 - 2 - 3 214 74

View Full Product Details