Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2D6
Entrez Gene ID
1565
Gene Name
cytochrome P450 family 2 subfamily D member 6
Gene Aliases
CPD6, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, P450-DB1, P450C2D, P450DB1
Chromosome Location
Chr.22: 42125531 - 42130881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025161.2 NP_001020332.2 2 - 3 440 85
XM_011529970.2 XP_011528272.1 2 - 3 434 85
GenBank mRNA AK309600.1 - 2 - 3 438 85
AY663390.1 - 1 - 2 174 85
BC066877.1 - 2 - 3 351 85
BC106757.1 - 2 - 3 372 85
CR456430.1 - 2 - 3 375 85

View Full Product Details