Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 76958901-76959412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACER3
Entrez Gene ID
55331
Gene Name
alkaline ceramidase 3
Gene Aliases
APHC, PHCA
UniGene
Hs.23862
Gene Chromosome Location
Chr.11: 76860873-77026797 on Build GRCh38
Transcripts
NM_001300953.1 NM_001300954.1 NM_001300955.1 NM_018367.6 XM_011545151.2 XM_011545152.2 XM_011545153.2 XM_017017987.1 XM_017017988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5027449 c.215-1G>A p.? Chr.11: 76958978-76958978 on Build GRCh38
7095847 c.259G>A p.E87K Chr.11: 76959023-76959023 on Build GRCh38
5645130 c.231C>T p.S77S Chr.11: 76958995-76958995 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs527691104 Chr.11: 76959375-76959375 on Build GRCh38
rs1013028141 Chr.11: 76959201-76959201 on Build GRCh38
rs113083206 Chr.11: 76959052-76959052 on Build GRCh38
rs567963089 Chr.11: 76959326-76959326 on Build GRCh38
rs914942793 Chr.11: 76959221-76959221 on Build GRCh38
rs551928333 Chr.11: 76959231-76959231 on Build GRCh38
rs548977140 Chr.11: 76959085-76959085 on Build GRCh38
rs142135871 Chr.11: 76959072-76959072 on Build GRCh38
rs781342423 Chr.11: 76958929-76958929 on Build GRCh38
rs537737531 Chr.11: 76959259-76959259 on Build GRCh38
rs889796595 Chr.11: 76959232-76959232 on Build GRCh38
rs752001543 Chr.11: 76959102-76959102 on Build GRCh38
rs904736378 Chr.11: 76959233-76959233 on Build GRCh38
rs937652325 Chr.11: 76959185-76959185 on Build GRCh38
rs1025752540 Chr.11: 76959310-76959310 on Build GRCh38
rs746011232 Chr.11: 76959139-76959139 on Build GRCh38
rs909793211 Chr.11: 76959032-76959032 on Build GRCh38
rs1042242935 Chr.11: 76959223-76959223 on Build GRCh38
rs776582624 Chr.11: 76959219-76959219 on Build GRCh38
rs758828041 Chr.11: 76959092-76959092 on Build GRCh38
rs757783921 Chr.11: 76959109-76959109 on Build GRCh38
rs11237033 Chr.11: 76959237-76959237 on Build GRCh38
rs972866216 Chr.11: 76959055-76959055 on Build GRCh38
rs766872615 Chr.11: 76959049-76959049 on Build GRCh38
rs971506525 Chr.11: 76959334-76959334 on Build GRCh38
rs186679888 Chr.11: 76959067-76959067 on Build GRCh38
rs191546675 Chr.11: 76958939-76958939 on Build GRCh38
rs781089214 Chr.11: 76959133-76959133 on Build GRCh38
rs537309536 Chr.11: 76959224-76959224 on Build GRCh38
rs774489309 Chr.11: 76959007-76959007 on Build GRCh38
rs567579035 Chr.11: 76959258-76959258 on Build GRCh38
rs753410842 Chr.11: 76959271-76959271 on Build GRCh38
rs182819789 Chr.11: 76959065-76959065 on Build GRCh38
rs377531957 Chr.11: 76958954-76958954 on Build GRCh38
rs764566631 Chr.11: 76959101-76959101 on Build GRCh38
rs545152313 Chr.11: 76958933-76958933 on Build GRCh38
rs761856919 Chr.11: 76959041-76959041 on Build GRCh38
rs375977493 Chr.11: 76958963-76958963 on Build GRCh38
rs745564093 Chr.11: 76959135-76959136 on Build GRCh38
rs186697519 Chr.11: 76958932-76958932 on Build GRCh38
rs1050979464 Chr.11: 76959229-76959229 on Build GRCh38
rs927667024 Chr.11: 76959138-76959138 on Build GRCh38
rs778377470 Chr.11: 76958954-76958954 on Build GRCh38
rs528601406 Chr.11: 76959227-76959227 on Build GRCh38
rs896025444 Chr.11: 76959166-76959166 on Build GRCh38
rs551339057 Chr.11: 76959273-76959273 on Build GRCh38
rs946606378 Chr.11: 76959222-76959222 on Build GRCh38
rs530818806 Chr.11: 76958989-76958989 on Build GRCh38
rs560082981 Chr.11: 76959225-76959225 on Build GRCh38

View Full Product Details