Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 83459174-83459679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLG2
Entrez Gene ID
1740
Gene Name
discs large MAGUK scaffold protein 2
Gene Aliases
PPP1R58, PSD-93, PSD93, chapsyn-110
UniGene
Hs.367656
Gene Chromosome Location
Chr.11: 83455009-85628534 on Build GRCh38
Transcripts
NM_001142699.1 NM_001142700.1 NM_001142702.1 NM_001206769.1 NM_001300983.1 NM_001364.3 XM_005273810.4 XM_005273811.4 XM_011544778.2 XM_011544780.2 XM_011544782.2 XM_011544788.2 XM_017017254.1 XM_017017255.1 XM_017017256.1 XM_017017257.1 XM_017017258.1 XM_017017259.1 XM_017017260.1 XM_017017261.1 XM_017017262.1 XM_017017263.1 XM_017017264.1 XM_017017265.1 XM_017017266.1 XM_017017267.1 XM_017017268.1 XM_017017269.1 XM_017017270.1 XM_017017271.1 XM_017017272.1 XM_017017273.1 XM_017017274.1 XM_017017275.1 XM_017017276.1 XM_017017277.1 XM_017017278.1 XM_017017279.1 XM_017017280.1 XM_017017281.1 XM_017017283.1 XM_017017284.1 XM_017017285.1 XM_017017286.1 XM_017017287.1 XM_017017288.1 XM_017017289.1 XM_017017290.1 XM_017017291.1 XM_017017292.1

SNP Detail

SNP ID Chromosome Location
rs150120827 Chr.11: 83459391-83459391 on Build GRCh38
rs887117982 Chr.11: 83459524-83459524 on Build GRCh38
rs1004387460 Chr.11: 83459544-83459544 on Build GRCh38
rs574737968 Chr.11: 83459559-83459559 on Build GRCh38
rs1006650827 Chr.11: 83459231-83459231 on Build GRCh38
rs141693822 Chr.11: 83459322-83459322 on Build GRCh38
rs771178762 Chr.11: 83459311-83459311 on Build GRCh38
rs1040792401 Chr.11: 83459535-83459535 on Build GRCh38
rs899899565 Chr.11: 83459488-83459488 on Build GRCh38
rs543051691 Chr.11: 83459560-83459560 on Build GRCh38
rs554913737 Chr.11: 83459639-83459639 on Build GRCh38
rs1017133051 Chr.11: 83459548-83459548 on Build GRCh38
rs1053406949 Chr.11: 83459212-83459212 on Build GRCh38
rs960714084 Chr.11: 83459641-83459641 on Build GRCh38
rs934102555 Chr.11: 83459494-83459494 on Build GRCh38
rs147351079 Chr.11: 83459486-83459486 on Build GRCh38
rs368905367 Chr.11: 83459654-83459654 on Build GRCh38
rs949211447 Chr.11: 83459443-83459443 on Build GRCh38
rs1002315095 Chr.11: 83459577-83459577 on Build GRCh38
rs980939801 Chr.11: 83459291-83459291 on Build GRCh38
rs142718943 Chr.11: 83459456-83459456 on Build GRCh38
rs377546939 Chr.11: 83459568-83459568 on Build GRCh38
rs778558419 Chr.11: 83459457-83459462 on Build GRCh38
rs145805160 Chr.11: 83459206-83459206 on Build GRCh38
rs926347514 Chr.11: 83459307-83459307 on Build GRCh38
rs889463590 Chr.11: 83459513-83459515 on Build GRCh38
rs900947034 Chr.11: 83459557-83459557 on Build GRCh38
rs546180181 Chr.11: 83459371-83459371 on Build GRCh38
rs770280940 Chr.11: 83459398-83459398 on Build GRCh38
rs148130918 Chr.11: 83459230-83459230 on Build GRCh38
rs138433454 Chr.11: 83459432-83459432 on Build GRCh38
rs1020164175 Chr.11: 83459233-83459233 on Build GRCh38
rs867685816 Chr.11: 83459327-83459327 on Build GRCh38
rs925227525 Chr.11: 83459425-83459425 on Build GRCh38
rs959008223 Chr.11: 83459396-83459396 on Build GRCh38
rs952964521 Chr.11: 83459336-83459336 on Build GRCh38
rs771615825 Chr.11: 83459633-83459633 on Build GRCh38
rs965573258 Chr.11: 83459293-83459293 on Build GRCh38
rs1033866927 Chr.11: 83459627-83459627 on Build GRCh38
rs992928596 Chr.11: 83459402-83459402 on Build GRCh38
rs4533060 Chr.11: 83459312-83459312 on Build GRCh38
rs1043673538 Chr.11: 83459410-83459410 on Build GRCh38
rs1051511453 Chr.11: 83459507-83459507 on Build GRCh38

View Full Product Details