Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112148477-112149011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18
Entrez Gene ID
3606
Gene Name
interleukin 18
Gene Aliases
IGIF, IL-18, IL-1g, IL1F4
UniGene
Hs.83077
Gene Chromosome Location
Chr.11: 112143251-112164117 on Build GRCh38
Transcripts
NM_001243211.1 NM_001562.3 XM_011542805.1 XM_011542806.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1351281 c.328C>A p.L110I Chr.11: 112148635-112148635 on Build GRCh38
4963554 c.258T>C p.S86S Chr.11: 112148705-112148705 on Build GRCh38
6257140 c.242C>A p.T81N Chr.11: 112148721-112148721 on Build GRCh38
5498583 c.330C>T p.L110L Chr.11: 112148633-112148633 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs34229190 Chr.11: 112148605-112148606 on Build GRCh38
rs757366374 Chr.11: 112148677-112148677 on Build GRCh38
rs576409374 Chr.11: 112148974-112148974 on Build GRCh38
rs45497197 Chr.11: 112148687-112148687 on Build GRCh38
rs988121717 Chr.11: 112148961-112148961 on Build GRCh38
rs397690770 Chr.11: 112148983-112148984 on Build GRCh38
rs770497566 Chr.11: 112148765-112148765 on Build GRCh38
rs961001177 Chr.11: 112148932-112148932 on Build GRCh38
rs768843343 Chr.11: 112148724-112148724 on Build GRCh38
rs149467995 Chr.11: 112148981-112148982 on Build GRCh38
rs981397741 Chr.11: 112148884-112148884 on Build GRCh38
rs10642361 Chr.11: 112148982-112148983 on Build GRCh38
rs1036370369 Chr.11: 112148827-112148827 on Build GRCh38
rs201878660 Chr.11: 112148595-112148595 on Build GRCh38
rs988889641 Chr.11: 112148918-112148918 on Build GRCh38
rs745920795 Chr.11: 112148685-112148685 on Build GRCh38
rs147751347 Chr.11: 112148782-112148782 on Build GRCh38
rs201058918 Chr.11: 112148725-112148725 on Build GRCh38
rs780531931 Chr.11: 112148706-112148706 on Build GRCh38
rs901538828 Chr.11: 112148691-112148691 on Build GRCh38
rs747430123 Chr.11: 112148717-112148717 on Build GRCh38
rs1043843985 Chr.11: 112148572-112148572 on Build GRCh38
rs1021334915 Chr.11: 112148951-112148951 on Build GRCh38
rs955704048 Chr.11: 112148883-112148883 on Build GRCh38
rs778806308 Chr.11: 112148684-112148684 on Build GRCh38
rs974540459 Chr.11: 112148973-112148973 on Build GRCh38
rs570059315 Chr.11: 112148609-112148609 on Build GRCh38
rs5744263 Chr.11: 112148818-112148818 on Build GRCh38
rs199696510 Chr.11: 112148748-112148748 on Build GRCh38
rs1037092169 Chr.11: 112148536-112148536 on Build GRCh38
rs536494545 Chr.11: 112148958-112148958 on Build GRCh38
rs201248108 Chr.11: 112148575-112148575 on Build GRCh38
rs141025779 Chr.11: 112148796-112148796 on Build GRCh38
rs1022834378 Chr.11: 112148798-112148798 on Build GRCh38
rs200345831 Chr.11: 112148776-112148776 on Build GRCh38
rs201399018 Chr.11: 112148836-112148836 on Build GRCh38
rs552834311 Chr.11: 112148938-112148938 on Build GRCh38
rs200101331 Chr.11: 112148983-112148983 on Build GRCh38
rs556495925 Chr.11: 112148963-112148963 on Build GRCh38
rs181711296 Chr.11: 112148929-112148929 on Build GRCh38
rs188899176 Chr.11: 112148848-112148848 on Build GRCh38
rs370406754 Chr.11: 112148756-112148756 on Build GRCh38
rs749416906 Chr.11: 112148675-112148675 on Build GRCh38
rs867479567 Chr.11: 112148689-112148689 on Build GRCh38
rs772922221 Chr.11: 112148749-112148749 on Build GRCh38
rs541144166 Chr.11: 112148528-112148528 on Build GRCh38
rs1004642225 Chr.11: 112148511-112148511 on Build GRCh38

View Full Product Details