Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 126303673-126304182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCPS
Entrez Gene ID
28960
Gene Name
decapping enzyme, scavenger
Gene Aliases
ARS, DCS1, HINT-5, HINT5, HSL1, HSPC015
UniGene
Hs.504249
Gene Chromosome Location
Chr.11: 126303301-126345754 on Build GRCh38
Transcripts
NM_014026.4 XM_011542778.2 XM_017017612.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
924680 c.52G>T p.E18* Chr.11: 126304132-126304132 on Build GRCh38
4574157 c.74C>A p.T25K Chr.11: 126304154-126304154 on Build GRCh38
5360598 c.16C>G p.P6A Chr.11: 126304096-126304096 on Build GRCh38
1627859 c.49G>T p.V17L Chr.11: 126304129-126304129 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1042114217 Chr.11: 126303958-126303958 on Build GRCh38
rs374596811 Chr.11: 126304119-126304119 on Build GRCh38
rs368061487 Chr.11: 126304045-126304045 on Build GRCh38
rs1034062720 Chr.11: 126304013-126304013 on Build GRCh38
rs141030550 Chr.11: 126304128-126304128 on Build GRCh38
rs374644970 Chr.11: 126304075-126304075 on Build GRCh38
rs933436874 Chr.11: 126303991-126303991 on Build GRCh38
rs927443449 Chr.11: 126304008-126304008 on Build GRCh38
rs902244421 Chr.11: 126303959-126303959 on Build GRCh38
rs542326802 Chr.11: 126303867-126303867 on Build GRCh38
rs760322945 Chr.11: 126304153-126304153 on Build GRCh38
rs371904995 Chr.11: 126304088-126304088 on Build GRCh38
rs3740915 Chr.11: 126304143-126304143 on Build GRCh38
rs1006898155 Chr.11: 126303746-126303747 on Build GRCh38
rs879363540 Chr.11: 126303972-126303972 on Build GRCh38
rs760319006 Chr.11: 126303848-126303848 on Build GRCh38
rs781363407 Chr.11: 126304124-126304124 on Build GRCh38
rs920566423 Chr.11: 126303716-126303716 on Build GRCh38
rs971442544 Chr.11: 126304000-126304000 on Build GRCh38
rs774065927 Chr.11: 126304139-126304139 on Build GRCh38
rs946289421 Chr.11: 126304134-126304134 on Build GRCh38
rs567395425 Chr.11: 126303943-126303943 on Build GRCh38
rs762311712 Chr.11: 126304050-126304050 on Build GRCh38
rs990033446 Chr.11: 126304042-126304042 on Build GRCh38
rs368512440 Chr.11: 126304081-126304081 on Build GRCh38
rs147755159 Chr.11: 126304097-126304097 on Build GRCh38
rs990474353 Chr.11: 126303975-126303975 on Build GRCh38
rs546055386 Chr.11: 126303752-126303752 on Build GRCh38
rs755244125 Chr.11: 126304122-126304122 on Build GRCh38
rs898112890 Chr.11: 126303997-126304002 on Build GRCh38
rs1041739415 Chr.11: 126303982-126303982 on Build GRCh38
rs958792932 Chr.11: 126304019-126304019 on Build GRCh38
rs1054732185 Chr.11: 126303895-126303895 on Build GRCh38
rs751282929 Chr.11: 126304067-126304067 on Build GRCh38
rs946314388 Chr.11: 126304035-126304035 on Build GRCh38
rs1018811477 Chr.11: 126303696-126303696 on Build GRCh38
rs201982035 Chr.11: 126304073-126304073 on Build GRCh38
rs112312647 Chr.11: 126303865-126303865 on Build GRCh38
rs759639289 Chr.11: 126304158-126304158 on Build GRCh38
rs971178265 Chr.11: 126303928-126303928 on Build GRCh38
rs1010563782 Chr.11: 126303941-126303941 on Build GRCh38
rs758435874 Chr.11: 126304086-126304086 on Build GRCh38
rs907905361 Chr.11: 126303994-126303994 on Build GRCh38
rs937562057 Chr.11: 126303878-126303878 on Build GRCh38
rs371807855 Chr.11: 126304066-126304066 on Build GRCh38
rs553765784 Chr.11: 126304127-126304127 on Build GRCh38
rs774656105 Chr.11: 126304101-126304103 on Build GRCh38
rs781602614 Chr.11: 126304096-126304096 on Build GRCh38
rs765873258 Chr.11: 126304065-126304065 on Build GRCh38
rs201108066 Chr.11: 126304117-126304117 on Build GRCh38
rs763469702 Chr.11: 126304105-126304105 on Build GRCh38
rs754624738 Chr.11: 126304072-126304072 on Build GRCh38
rs972359904 Chr.11: 126303871-126303871 on Build GRCh38
rs538949365 Chr.11: 126304012-126304012 on Build GRCh38
rs990314891 Chr.11: 126304126-126304126 on Build GRCh38
rs770582008 Chr.11: 126304106-126304106 on Build GRCh38
rs558922669 Chr.11: 126304064-126304064 on Build GRCh38
rs914825239 Chr.11: 126304032-126304032 on Build GRCh38
rs566444713 Chr.11: 126303985-126303985 on Build GRCh38
rs767262030 Chr.11: 126304110-126304110 on Build GRCh38
rs944359568 Chr.11: 126304155-126304155 on Build GRCh38
rs531961986 Chr.11: 126303826-126303826 on Build GRCh38
rs769030662 Chr.11: 126304039-126304039 on Build GRCh38
rs907959555 Chr.11: 126303793-126303793 on Build GRCh38
rs1008678492 Chr.11: 126303966-126303966 on Build GRCh38
rs889174053 Chr.11: 126304091-126304091 on Build GRCh38
rs34534864 Chr.11: 126304087-126304087 on Build GRCh38
rs143223999 Chr.11: 126303983-126303983 on Build GRCh38
rs927495917 Chr.11: 126303876-126303876 on Build GRCh38
rs538087473 Chr.11: 126304107-126304107 on Build GRCh38
rs554430727 Chr.11: 126304113-126304113 on Build GRCh38
rs180849635 Chr.11: 126303927-126303927 on Build GRCh38
rs1050474343 Chr.11: 126304090-126304090 on Build GRCh38
rs1055067495 Chr.11: 126304031-126304031 on Build GRCh38
rs143814193 Chr.11: 126303981-126303981 on Build GRCh38
rs914387266 Chr.11: 126304058-126304058 on Build GRCh38
rs1034935205 Chr.11: 126303812-126303812 on Build GRCh38
rs772829450 Chr.11: 126304060-126304060 on Build GRCh38
rs1018758651 Chr.11: 126303970-126303970 on Build GRCh38
rs1025308899 Chr.11: 126303896-126303896 on Build GRCh38
rs540634001 Chr.11: 126303699-126303699 on Build GRCh38
rs35061643 Chr.11: 126303708-126303708 on Build GRCh38
rs752586869 Chr.11: 126304111-126304111 on Build GRCh38
rs923559735 Chr.11: 126304150-126304150 on Build GRCh38
rs959082089 Chr.11: 126304125-126304125 on Build GRCh38
rs750552277 Chr.11: 126304082-126304082 on Build GRCh38
rs889277421 Chr.11: 126303742-126303742 on Build GRCh38
rs993749916 Chr.11: 126303999-126304001 on Build GRCh38
rs34631616 Chr.11: 126303714-126303714 on Build GRCh38
rs555806352 Chr.11: 126304002-126304002 on Build GRCh38
rs983909435 Chr.11: 126303763-126303763 on Build GRCh38
rs775032697 Chr.11: 126304147-126304147 on Build GRCh38
rs952084192 Chr.11: 126303759-126303759 on Build GRCh38
rs763808337 Chr.11: 126304154-126304154 on Build GRCh38
rs562827071 Chr.11: 126303778-126303778 on Build GRCh38
rs962695747 Chr.11: 126303859-126303859 on Build GRCh38
rs914460529 Chr.11: 126303980-126303980 on Build GRCh38
rs773623495 Chr.11: 126304104-126304104 on Build GRCh38
rs1045859288 Chr.11: 126303866-126303866 on Build GRCh38
rs543115950 Chr.11: 126303955-126303955 on Build GRCh38
rs771563009 Chr.11: 126304138-126304138 on Build GRCh38
rs752539882 Chr.11: 126304076-126304076 on Build GRCh38
rs765650762 Chr.11: 126304054-126304054 on Build GRCh38
rs1008625825 Chr.11: 126304074-126304074 on Build GRCh38
rs756132248 Chr.11: 126304080-126304080 on Build GRCh38
rs760352546 Chr.11: 126304112-126304112 on Build GRCh38
rs776113371 Chr.11: 126304131-126304131 on Build GRCh38
rs981452494 Chr.11: 126304120-126304120 on Build GRCh38
rs759331827 Chr.11: 126304108-126304108 on Build GRCh38
rs759091675 Chr.11: 126304141-126304141 on Build GRCh38
rs964555819 Chr.11: 126303971-126303971 on Build GRCh38
rs749768260 Chr.11: 126304136-126304136 on Build GRCh38
rs376410163 Chr.11: 126304121-126304121 on Build GRCh38
rs983469738 Chr.11: 126303992-126303992 on Build GRCh38

View Full Product Details