Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1806167-1806671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB1
Entrez Gene ID
2782
Gene Name
G protein subunit beta 1
Gene Aliases
MRD42
UniGene
Hs.430425
Gene Chromosome Location
Chr.1: 1785285-1891117 on Build GRCh38
Transcripts
NM_001282538.1 NM_001282539.1 NM_002074.4 XM_017001059.1 XM_017001060.1 XM_017001061.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4444237 c.225G>T p.Q75H Chr.1: 1806517-1806517 on Build GRCh38
1290046 c.239T>C p.I80T Chr.1: 1806503-1806503 on Build GRCh38
4970075 c.266A>C p.K89T Chr.1: 1806476-1806476 on Build GRCh38
899791 c.222G>A p.S74S Chr.1: 1806520-1806520 on Build GRCh38
4994175 c.265A>G p.K89E Chr.1: 1806477-1806477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759614261 Chr.1: 1806464-1806464 on Build GRCh38
rs869312821 Chr.1: 1806515-1806515 on Build GRCh38
rs903427757 Chr.1: 1806310-1806310 on Build GRCh38
rs370522649 Chr.1: 1806437-1806437 on Build GRCh38
rs756962211 Chr.1: 1806279-1806279 on Build GRCh38
rs769640777 Chr.1: 1806544-1806544 on Build GRCh38
rs999560777 Chr.1: 1806287-1806287 on Build GRCh38
rs752746786 Chr.1: 1806503-1806503 on Build GRCh38
rs869312823 Chr.1: 1806509-1806509 on Build GRCh38
rs191914515 Chr.1: 1806435-1806435 on Build GRCh38
rs1040371792 Chr.1: 1806212-1806212 on Build GRCh38
rs909938942 Chr.1: 1806563-1806563 on Build GRCh38
rs776589515 Chr.1: 1806428-1806428 on Build GRCh38
rs750375782 Chr.1: 1806443-1806443 on Build GRCh38
rs77354509 Chr.1: 1806456-1806456 on Build GRCh38
rs984519325 Chr.1: 1806239-1806239 on Build GRCh38
rs570008358 Chr.1: 1806396-1806396 on Build GRCh38
rs774015213 Chr.1: 1806572-1806572 on Build GRCh38
rs545209841 Chr.1: 1806520-1806520 on Build GRCh38
rs546898244 Chr.1: 1806619-1806619 on Build GRCh38
rs143096846 Chr.1: 1806255-1806255 on Build GRCh38
rs767940181 Chr.1: 1806457-1806457 on Build GRCh38
rs557812266 Chr.1: 1806251-1806251 on Build GRCh38
rs765296776 Chr.1: 1806552-1806553 on Build GRCh38
rs747011357 Chr.1: 1806430-1806430 on Build GRCh38
rs936226961 Chr.1: 1806606-1806606 on Build GRCh38
rs1054064055 Chr.1: 1806616-1806616 on Build GRCh38
rs934137563 Chr.1: 1806294-1806294 on Build GRCh38
rs768214138 Chr.1: 1806565-1806565 on Build GRCh38
rs748794833 Chr.1: 1806574-1806574 on Build GRCh38
rs758432471 Chr.1: 1806513-1806513 on Build GRCh38
rs557288106 Chr.1: 1806324-1806324 on Build GRCh38
rs993733911 Chr.1: 1806442-1806442 on Build GRCh38
rs770756474 Chr.1: 1806433-1806433 on Build GRCh38
rs372291632 Chr.1: 1806585-1806585 on Build GRCh38
rs74898646 Chr.1: 1806455-1806455 on Build GRCh38
rs771178392 Chr.1: 1806427-1806428 on Build GRCh38
rs149782013 Chr.1: 1806532-1806532 on Build GRCh38
rs571247703 Chr.1: 1806252-1806252 on Build GRCh38
rs761262096 Chr.1: 1806582-1806582 on Build GRCh38
rs768250415 Chr.1: 1806452-1806452 on Build GRCh38
rs869312822 Chr.1: 1806514-1806514 on Build GRCh38
rs781140490 Chr.1: 1806434-1806434 on Build GRCh38
rs573831548 Chr.1: 1806334-1806334 on Build GRCh38
rs761135568 Chr.1: 1806465-1806465 on Build GRCh38
rs1016919382 Chr.1: 1806493-1806493 on Build GRCh38
rs894173154 Chr.1: 1806618-1806618 on Build GRCh38
rs17363334 Chr.1: 1806461-1806461 on Build GRCh38
rs984139062 Chr.1: 1806481-1806481 on Build GRCh38
rs1042345377 Chr.1: 1806638-1806638 on Build GRCh38
rs375820150 Chr.1: 1806545-1806545 on Build GRCh38
rs1054012132 Chr.1: 1806366-1806366 on Build GRCh38
rs561258180 Chr.1: 1806590-1806590 on Build GRCh38
rs761234962 Chr.1: 1806573-1806573 on Build GRCh38
rs766838810 Chr.1: 1806478-1806478 on Build GRCh38
rs934302044 Chr.1: 1806620-1806620 on Build GRCh38
rs903351766 Chr.1: 1806286-1806286 on Build GRCh38
rs748987967 Chr.1: 1806552-1806552 on Build GRCh38
rs372953414 Chr.1: 1806499-1806499 on Build GRCh38
rs530382280 Chr.1: 1806613-1806613 on Build GRCh38
rs1020487478 Chr.1: 1806282-1806282 on Build GRCh38
rs187619840 Chr.1: 1806388-1806388 on Build GRCh38
rs755167846 Chr.1: 1806425-1806425 on Build GRCh38
rs894248099 Chr.1: 1806389-1806389 on Build GRCh38
rs907058034 Chr.1: 1806436-1806436 on Build GRCh38
rs768589187 Chr.1: 1806460-1806461 on Build GRCh38
rs553106192 Chr.1: 1806393-1806393 on Build GRCh38
rs762586996 Chr.1: 1806448-1806448 on Build GRCh38
rs1042013065 Chr.1: 1806296-1806296 on Build GRCh38
rs901399270 Chr.1: 1806240-1806240 on Build GRCh38
rs754181534 Chr.1: 1806487-1806487 on Build GRCh38

View Full Product Details