Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14775343-14775848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf60
Entrez Gene ID
144608
Gene Name
chromosome 12 open reading frame 60
Gene Aliases
-
UniGene
Hs.659951
Gene Chromosome Location
Chr.12: 14567991-14823858 on Build GRCh38
Transcripts
XM_017018871.1 XM_017018872.1 NM_177925.3 NR_027716.2

Gene Symbol
H2AFJ
Entrez Gene ID
55766
Gene Name
H2A histone family member J
Gene Aliases
H2AJ
UniGene
Hs.524280
Gene Chromosome Location
Chr.12: 14774336-14778002 on Build GRCh38
Transcripts
XM_017018871.1 XM_017018872.1 NM_177925.3 NR_027716.2

SNP Detail

SNP ID Chromosome Location
rs1049194539 Chr.12: 14775421-14775421 on Build GRCh38
rs939958274 Chr.12: 14775591-14775591 on Build GRCh38
rs889317192 Chr.12: 14775435-14775435 on Build GRCh38
rs1020972248 Chr.12: 14775534-14775534 on Build GRCh38
rs566811865 Chr.12: 14775379-14775379 on Build GRCh38
rs1009548538 Chr.12: 14775530-14775530 on Build GRCh38
rs904117856 Chr.12: 14775580-14775580 on Build GRCh38
rs147529853 Chr.12: 14775683-14775683 on Build GRCh38
rs1015905546 Chr.12: 14775717-14775717 on Build GRCh38
rs995219343 Chr.12: 14775419-14775419 on Build GRCh38
rs932399497 Chr.12: 14775801-14775801 on Build GRCh38
rs983487465 Chr.12: 14775685-14775685 on Build GRCh38
rs367739651 Chr.12: 14775818-14775818 on Build GRCh38
rs561238110 Chr.12: 14775622-14775622 on Build GRCh38
rs866384099 Chr.12: 14775597-14775597 on Build GRCh38
rs528809078 Chr.12: 14775628-14775628 on Build GRCh38
rs972332670 Chr.12: 14775626-14775626 on Build GRCh38
rs1001423813 Chr.12: 14775589-14775589 on Build GRCh38
rs748705980 Chr.12: 14775689-14775689 on Build GRCh38
rs183316337 Chr.12: 14775736-14775736 on Build GRCh38
rs979959942 Chr.12: 14775536-14775536 on Build GRCh38
rs575769454 Chr.12: 14775592-14775592 on Build GRCh38
rs35291781 Chr.12: 14775398-14775398 on Build GRCh38
rs375110022 Chr.12: 14775667-14775667 on Build GRCh38
rs138878306 Chr.12: 14775392-14775392 on Build GRCh38
rs761246842 Chr.12: 14775416-14775416 on Build GRCh38
rs902110354 Chr.12: 14775462-14775462 on Build GRCh38
rs1007792187 Chr.12: 14775439-14775439 on Build GRCh38
rs962679395 Chr.12: 14775734-14775734 on Build GRCh38
rs553335074 Chr.12: 14775501-14775501 on Build GRCh38
rs11056184 Chr.12: 14775636-14775636 on Build GRCh38
rs1033876394 Chr.12: 14775599-14775599 on Build GRCh38
rs139499522 Chr.12: 14775537-14775537 on Build GRCh38
rs945063015 Chr.12: 14775554-14775554 on Build GRCh38
rs1050708027 Chr.12: 14775453-14775453 on Build GRCh38
rs746615189 Chr.12: 14775383-14775383 on Build GRCh38
rs921129210 Chr.12: 14775759-14775759 on Build GRCh38
rs542845900 Chr.12: 14775604-14775604 on Build GRCh38
rs919600425 Chr.12: 14775668-14775670 on Build GRCh38
rs917571456 Chr.12: 14775374-14775374 on Build GRCh38
rs545497317 Chr.12: 14775548-14775548 on Build GRCh38
rs1049967889 Chr.12: 14775732-14775732 on Build GRCh38
rs746009601 Chr.12: 14775448-14775448 on Build GRCh38
rs757688536 Chr.12: 14775712-14775712 on Build GRCh38
rs1013717507 Chr.12: 14775645-14775645 on Build GRCh38
rs890830674 Chr.12: 14775457-14775457 on Build GRCh38
rs968092196 Chr.12: 14775536-14775536 on Build GRCh38
rs1019643735 Chr.12: 14775446-14775446 on Build GRCh38
rs754515313 Chr.12: 14775514-14775514 on Build GRCh38
rs932430445 Chr.12: 14775418-14775418 on Build GRCh38
rs544558417 Chr.12: 14775451-14775451 on Build GRCh38
rs578003788 Chr.12: 14775512-14775512 on Build GRCh38
rs1036667343 Chr.12: 14775381-14775381 on Build GRCh38
rs781266559 Chr.12: 14775415-14775415 on Build GRCh38
rs754328727 Chr.12: 14775367-14775367 on Build GRCh38
rs767404752 Chr.12: 14775607-14775607 on Build GRCh38
rs17761740 Chr.12: 14775559-14775559 on Build GRCh38
rs191987045 Chr.12: 14775686-14775686 on Build GRCh38
rs750128910 Chr.12: 14775752-14775752 on Build GRCh38
rs898226253 Chr.12: 14775389-14775389 on Build GRCh38

View Full Product Details