Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 21508273-21508817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLT1B
Entrez Gene ID
51026
Gene Name
golgi transport 1B
Gene Aliases
CGI-141, GCT2, GOT1, GOT1B, YMR292W
UniGene
Hs.62275
Gene Chromosome Location
Chr.12: 21501765-21518403 on Build GRCh38
Transcripts
NM_016072.4 XM_017019384.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6891940 c.283T>A p.F95I Chr.12: 21508548-21508548 on Build GRCh38
1685687 c.207delT p.L72fs*6 Chr.12: 21508472-21508472 on Build GRCh38
692805 c.256G>T p.G86C Chr.12: 21508521-21508521 on Build GRCh38
2005142 c.201T>C p.A67A Chr.12: 21508466-21508466 on Build GRCh38
281428 c.174C>A p.F58L Chr.12: 21508439-21508439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377112868 Chr.12: 21508515-21508515 on Build GRCh38
rs773517026 Chr.12: 21508580-21508583 on Build GRCh38
rs769552575 Chr.12: 21508553-21508553 on Build GRCh38
rs769778292 Chr.12: 21508391-21508391 on Build GRCh38
rs549420247 Chr.12: 21508524-21508524 on Build GRCh38
rs745400758 Chr.12: 21508576-21508576 on Build GRCh38
rs979947872 Chr.12: 21508335-21508335 on Build GRCh38
rs775525196 Chr.12: 21508598-21508598 on Build GRCh38
rs945451950 Chr.12: 21508455-21508455 on Build GRCh38
rs763937560 Chr.12: 21508428-21508428 on Build GRCh38
rs887907221 Chr.12: 21508539-21508539 on Build GRCh38
rs907013051 Chr.12: 21508778-21508778 on Build GRCh38
rs150036114 Chr.12: 21508567-21508567 on Build GRCh38
rs756080215 Chr.12: 21508475-21508475 on Build GRCh38
rs755446813 Chr.12: 21508541-21508541 on Build GRCh38
rs749668605 Chr.12: 21508357-21508357 on Build GRCh38
rs768688915 Chr.12: 21508608-21508608 on Build GRCh38
rs563895527 Chr.12: 21508321-21508321 on Build GRCh38
rs762173690 Chr.12: 21508458-21508458 on Build GRCh38
rs372982409 Chr.12: 21508451-21508451 on Build GRCh38
rs751366542 Chr.12: 21508457-21508457 on Build GRCh38
rs386761078 Chr.12: 21508708-21508710 on Build GRCh38
rs1038149496 Chr.12: 21508680-21508680 on Build GRCh38
rs11046093 Chr.12: 21508773-21508773 on Build GRCh38
rs762835460 Chr.12: 21508601-21508601 on Build GRCh38
rs548752443 Chr.12: 21508616-21508616 on Build GRCh38
rs530194085 Chr.12: 21508758-21508758 on Build GRCh38
rs11046092 Chr.12: 21508708-21508708 on Build GRCh38
rs200318646 Chr.12: 21508383-21508383 on Build GRCh38
rs780623728 Chr.12: 21508507-21508507 on Build GRCh38
rs915293344 Chr.12: 21508432-21508432 on Build GRCh38
rs376744318 Chr.12: 21508336-21508336 on Build GRCh38
rs763383992 Chr.12: 21508590-21508591 on Build GRCh38
rs11046091 Chr.12: 21508574-21508574 on Build GRCh38
rs756701151 Chr.12: 21508341-21508341 on Build GRCh38
rs765484210 Chr.12: 21508370-21508370 on Build GRCh38
rs770435450 Chr.12: 21508582-21508583 on Build GRCh38
rs369058204 Chr.12: 21508351-21508351 on Build GRCh38
rs1049146374 Chr.12: 21508537-21508537 on Build GRCh38
rs775237194 Chr.12: 21508397-21508397 on Build GRCh38
rs777435736 Chr.12: 21508379-21508379 on Build GRCh38
rs762824979 Chr.12: 21508410-21508410 on Build GRCh38
rs776179457 Chr.12: 21508388-21508388 on Build GRCh38
rs941003310 Chr.12: 21508631-21508631 on Build GRCh38
rs769141819 Chr.12: 21508597-21508597 on Build GRCh38
rs746705153 Chr.12: 21508380-21508380 on Build GRCh38
rs767836925 Chr.12: 21508464-21508464 on Build GRCh38
rs750581064 Chr.12: 21508466-21508466 on Build GRCh38
rs61926330 Chr.12: 21508307-21508307 on Build GRCh38
rs889498024 Chr.12: 21508625-21508625 on Build GRCh38
rs768853899 Chr.12: 21508358-21508358 on Build GRCh38
rs547777319 Chr.12: 21508785-21508785 on Build GRCh38
rs550560510 Chr.12: 21508605-21508608 on Build GRCh38
rs779300870 Chr.12: 21508543-21508543 on Build GRCh38
rs986401869 Chr.12: 21508301-21508301 on Build GRCh38
rs896913615 Chr.12: 21508703-21508703 on Build GRCh38
rs866673013 Chr.12: 21508331-21508331 on Build GRCh38

View Full Product Details